EMAIL THIS PAGE TO A FRIEND

255572 Sigma-Aldrich

Potassium chlorate

ACS reagent, ≥99.0%

Purchase

Properties

Related Categories Chemical Synthesis, Essential Chemicals, Inorganic Salts, Potassium, Potassium Salts,
grade   ACS reagent
assay   ≥99.0%
impurities   ≤0.001% N compounds
  ≤0.005% insolubles
mp   356 °C(lit.)
anion traces   bromate (BrO3-): ≤0.015%
  chloride (Cl-): ≤0.001%
  sulfate (SO42-): passes test
cation traces   Ca: ≤0.002%
  Fe: ≤3 ppm
  Mg: ≤0.002%
  Na: ≤0.01%
  heavy metals (as Pb): ≤5 ppm

Description

Packaging

100, 500 g in poly bottle

Price and Availability


Ultra-high Purity Salts

Additional Products to Explore

Hydrochloric acid

ACS reagent, 37%

Potassium chloride

BioXtra, ≥99.0%

Potassium nitrate

ReagentPlus®, ≥99.0%

Potassium perchlorate

ACS reagent, ≥99%

Sulfuric acid

99.999%

Customers Also Viewed

Potassium chlorate

BioXtra, ≥99.0%

Potassium chlorate

puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%

Potassium chlorate

puriss., 99-101%

Potassium chlorate

purum p.a., crystallized, ≥99.0% (T)

Potassium perchlorate

ACS reagent, ≥99%

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Hazard Codes (Europe) 
Risk Statements (Europe) 
Safety Statements (Europe) 
13-16-27-61
RIDADR 
UN 1485 5.1 / PGII
WGK Germany 
2
RTECS 
FO0350000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Suicidal intoxication with potassium chlorate successfully treated with renal replacement therapy and extracorporeal liver support. Sein Anand J, Barwina M, Zajac M, et al. Prz. Lek. 69(8), 585-6, (2012)

Loading...


[Markers of chloric acid (I) in biological systems--identification and properties]. Robaszkiewicz A Postepy Biochem. 56(2), 201-8, (2010)

Loading...


Solubilizing properties of new surface-active agents, products of selective oxyethylation of cholic acid. Part I. Zgoda MM, Nachajski MJ, Kołodziejczyk MK, et al. Polim. Med. 37(4), 21-38, (2007)

Loading...


[Influence of gastric juice pH on the metabolism of vitamin C in gastric mucosa and juice]. Safranow K, Korzonek M, Dziedziejko V, et al. Pol. Merkur. Lekarski 20(116), 168-72, (2006)

Loading...


[Acid producing function of the stomach in patients with duodenal ulcer and diabetes mellitus]. Smolianinov AB, Novitskiĭ VA, and Lebedev NN Klin. Med. (Mosk.) 79(8), 44-7, (2001)

Loading...


Risk of upper gastrointestinal tract bleeding associated with selective serotonin reuptake inhibitors and venlafaxine therapy: interaction with nonsteroidal anti-inflammatory drugs and effect of acid-suppressing agents. de Abajo FJ and García-Rodríguez LA Arch. Gen. Psychiatry 65(7), 795-803, (2008)

Loading...


Prenatal exposure to the CB1 receptor agonist WIN 55,212-2 causes learning disruption associated with impaired cortical NMDA receptor function and emotional reactivity changes in rat offspring. Antonelli T, Tomasini MC, Tattoli M, et al. Cereb. Cortex 15(12), 2013-20, (2005)

Loading...


Alcohol-induced versus anion-induced states of alpha-chymotrypsinogen A at low pH. Khan F, Khan RH, and Muzammil S Biochim. Biophys. Acta 1481(2), 229-36, (2000)

Loading...


Electroactivity of a starburst hole-transport material in Langmuir-Blodgett films. Solid state effects and intervalence charge transfer. Parra V, Del Caño T, Rodríguez-Méndez ML, et al. Phys. Chem. Chem. Phys. 9(18), 2266-73, (2007)

Loading...


Development of a modified positive selection medium that allows to isolate Aspergillus oryzae strains cured of the integrated niaD-based plasmid. Ishi K, Watanabe T, Juvvadi PR, et al. Biosci. Biotechnol. Biochem. 69(12), 2463-5, (2005)

Loading...


Selective inhibition of nitrite oxidation by chlorate dosing in aerobic granules. Xu G, Xu X, Yang F, et al. J. Hazard. Mater. 185(1), 249-54, (2011)

Loading...


Evaluation of chlorine dioxide gas residues on selected food produce. Trinetta V, Vaidya N, Linton R, et al. J. Food Sci. 76(1), T11-5, (2011)

Loading...


Investigation of plant hormone level changes in shoot tips of longan (Dimocarpus longan Lour.) treated with potassium chlorate by liquid chromatography-electrospray ionization mass spectrometry. Susawaengsup C, Rayanakorn M, et al. Talanta 85, 897-905, (2011)

Loading...


Development of interdigitated array electrodes with surface-enhanced Raman scattering functionality. Islam MM, Ueno K, Juodkazis S, et al. Anal. Sci. 26(1), 13-8, (2010)

Loading...


Interaction of human chondrocytes and NIH/3T3 fibroblasts on chloric acid-treated biodegradable polymer surfaces. Lee SJ, Khang G, Lee YM, et al. J. Biomater. Sci. Polym. Ed. 13(2), 197-212, (2002)

Loading...


Thermal decomposition and isomerization of cis-permethrin and beta-cypermethrin in the solid phase. González Audino P, Licastro SA, and Zerba E Pest Manag. Sci. 58(2), 183-9, (2002)

Loading...


Mechanism of thrombin-induced vasodilation in human coronary arterioles. Bosnjak JJ, Terata K, Miura H, et al. Am. J. Physiol. Heart Circ. Physiol. 284(4), H1080-6, (2003)

Loading...


Cranial MR imaging findings of potassium chlorate intoxication. Mutlu H, Silit E, Pekkafali Z, et al. Am. J. Neuroradiol. 24(7), 1396-8, (2003)

Loading...


The role of pendrin in iodide regulation. Fugazzola L, Cerutti N, Mannavola D, et al. Exp. Clin. Endocrinol. Diabetes 109(1), 18-22, (2001)

Loading...


Numerical simulation of NQR/NMR: Applications in quantum computing. Possa D, Gaudio AC, and Freitas JC J. Magn. Reson. 209(2), 250-60, (2011)

Loading...


Chloric acid(I) affects antioxidant defense of lung epitelial cells. Robaszkiewicz A, Pogorzelska M, Bartosz G, et al. Toxicol. In Vitro 25(7), 1328-34, (2011)

Loading...


Determination of trace selenium by solid substrate-room temperature phosphorescence enhancing method based on potassium chlorate oxidizing phenyl hydrazine-1,2-dihydroxynaphthalene-3,6-disulfonic acid system. Liu JM, Cui XJ, Li LM, et al. Spectrochim. Acta. A. Mol. Biomol. Spectrosc. 66(4-5), 1194-8, (2007)

Loading...


Effect of drug particle size in ultrasound compacted tablets. Continuum percolation model approach. Millán M and Caraballo I Int. J. Pharm. 310(1-2), 168-74, (2006)

Loading...


Removal of 5-amino-2-chlorotoluene-4-sulfonic and chlorhydric acids from wastewater by weakly basic resin: equilibrium and kinetics. Li CH Biomed. Environ. Sci. 20(5), 392-7, (2007)

Loading...


Continuous infusion of calcium gluconate in 5% albumin is safe and prevents most hypocalcemic reactions during therapeutic plasma exchange. Kankirawatana S, Huang ST, and Marques MB J. Clin. Apher. 22(5), 265-9, (2007)

Loading...


Merck 14,7620

Fieser 1,927

Corp MSDS 1 (2), 2897:B / FT-IR 2 (3), 4761:B / FT-IR 1 (2), 1252:C / RegBook 1 (3), 3455:B / Sax 6, 2271

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?