Synonym: Hydroxylammonium chloride
CAS Number 5470-11-1
Linear Formula NH2OH · HCl
Molecular Weight 69.49
Beilstein Registry Number 3539763
EC Number 226-798-2
MDL number MFCD00051089
PubChem Substance ID 24855466
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, Bioactive Small Molecules, C-X Bond Formation (Non-Halogen), Cell Biology, Chemical Synthesis, |
| grade | ACS reagent |
| assay | 98.0% |
| impurities | ≤0.005% S compounds |
| ≤0.25 meq/g Titr. free acid | |
| ign. residue | ≤0.05% |
| mp | 155-157 °C (dec.)(lit.) |
| density | 1.67 g/mL at 25 °C(lit.) |
Powerful reducing agent. Converts aldehydes and ketones to oximes, and acid chlorides to hydroxamic acids. Used as catalyst, swelling agent, and copolymerization inhibitor.
MAO inhibitor; inhibits platelet aggregation.
100, 500 g in poly bottle
5 g in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?