• USA Home
  • 255599 - Cobalt(II) chloride hexahydrate

EMAIL THIS PAGE TO A FRIEND
255599 Sigma-Aldrich

Cobalt(II) chloride hexahydrate

ACS reagent, 98%

Synonym: Cobaltous chloride hexahydrate

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Cobalt, Cobalt Salts, Essential Chemicals,
Quality Level   PREMIUM
grade   ACS reagent
vapor pressure   40 mmHg ( 0 °C)
assay   98%
  98.0-102.0% (ACS specification)
impurities   ≤0.01% insolubles
anion traces   nitrate (NO3-): ≤0.01%
  sulfate (SO42-): ≤0.01%
cation traces   Ca: ≤0.005%
  Cu: ≤0.002%
  Fe: ≤0.005%
  K: ≤0.01%
  Mg: ≤0.005%
  Na: ≤0.05%
  Ni: ≤0.1%
  Zn: ≤0.03%

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Packaging

100, 500 g in poly bottle

5 g in glass bottle

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




Great Products. Great New Price!
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
GG0200000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?