• USA Home
  • 25610 - 2-Chloro-5-nitropyridine

EMAIL THIS PAGE TO A FRIEND
25610 Sigma-Aldrich

2-Chloro-5-nitropyridine

purum, ≥97.0% (AT)

  • CAS Number 4548-45-2

  • Empirical Formula (Hill Notation) C5H3ClN2O2

  • Molecular Weight 158.54

  •  Beilstein Registry Number 120453

  •  EC Number 224-908-3

  •  MDL number MFCD00006240

  •  PubChem Substance ID 329752802

Purchase

Properties

Related Categories Building Blocks, C5, Chemical Synthesis, Halogenated Heterocycles, Heterocyclic Building Blocks,
InChI Key   BAZVFQBTJPBRTJ-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (AT)
mp   105-108 °C(lit.)
  105-109 °C

Description

Caution

may discolor to a dark color on storage

Application

2-Chloro-5-nitropyridine has been used in preparation of 5,5′-dinitro-2,2′-dipyridylsulfide.

General description

Kinetics of the reaction of 2-chloro-5-nitropyridine with a series of aryloxide ions has been studied in methanol.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
US7175000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?