EMAIL THIS PAGE TO A FRIEND
25635 Sigma-Aldrich

1-Chlorononane

purum, ≥96.0% (GC)

Synonym: Nonyl chloride

  • CAS Number 2473-01-0

  • Linear Formula CH3(CH2)8Cl

  • Molecular Weight 162.70

  •  Beilstein Registry Number 1733625

  •  EC Number 219-595-5

  •  MDL number MFCD00001023

Purchase

Properties

InChI Key   RKAMCQVGHFRILV-UHFFFAOYSA-N
grade   purum
assay   ≥96.0% (GC)
refractive index   n20/D 1.436(lit.)
  n20/D 1.436
bp   202-204 °C(lit.)
density   0.87 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
165 °F
Flash Point(C) 
74 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

238457 1-Chlorononane, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?