EMAIL THIS PAGE TO A FRIEND
256420 Sigma-Aldrich

Trichloroethylene

spectrophotometric grade, ≥99.5%

Synonym: TCE, Trichloroethene

Purchase

Properties

Related Categories Amber Glass Bottles, Solvent Bottles, Solvent Packaging Options, Solvent by Application, Solvents,
grade   spectrophotometric grade
vapor density   4.5 (vs air)
vapor pressure   61 mmHg ( 20 °C)
InChI Key   XSTXAVWGXDQKEL-UHFFFAOYSA-N
assay   ≥99.5%
autoignition temp.   770 °F
impurities   <0.020% water
evapn. residue   <0.001%
refractive index   n20/D 1.476(lit.)
bp   86.7 °C(lit.)
mp   −84.8 °C(lit.)
density   1.463 g/mL at 25 °C(lit.)
λ   H2O reference
UV absorption   λ: 278 nm Amax: 1.00
  λ: 300 nm Amax: 0.10
  λ: 325 nm Amax: 0.08
  λ: 350-400 nm Amax: 0.06

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1710 6.1 / PGIII
WGK Germany 
3
RTECS 
KX4550000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?