• USA Home
  • 25745 - Chloroquine diphosphate salt

EMAIL THIS PAGE TO A FRIEND
25745 Sigma-Aldrich

Chloroquine diphosphate salt

≥96.0% (TLC)

Synonym: N4-(7-Chloro-4-quinolinyl)-N1,N1-dimethyl-1,4-pentanediamine diphosphate salt

  • CAS Number 50-63-5

  • Empirical Formula (Hill Notation) C18H26ClN3 · 2H3PO4

  • Molecular Weight 515.86

  •  Beilstein Registry Number 4223142

  •  EC Number 200-055-2

  •  MDL number MFCD00069852

Purchase

Properties

InChI Key   QKICWELGRMTQCR-UHFFFAOYSA-N
assay   ≥96.0% (TLC)
impurities   ≤3% water
mp   200 °C (dec.)(lit.)
solubility   H2O: 50 mg/mL clear
Mode of action   enzyme | inhibits

Description

Other Notes

Lysosomotropic drug

This product has been replaced by C6628-SIGMA | Chloroquine diphosphate salt solid, ≥98%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
VB2450000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C6628 Chloroquine diphosphate salt, solid, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?