• USA Home
  • 25865 - 4-Chlorophenol solution

EMAIL THIS PAGE TO A FRIEND
25865 Sigma-Aldrich

4-Chlorophenol solution

725 mg/L in H2O, for AOX determination (according to DIN 38409-H14)

  • CAS Number 106-48-9

  • Empirical Formula (Hill Notation) C6H5ClO

  • Molecular Weight 128.56

  •  Beilstein Registry Number 507004

  •  MDL number MFCD00002318

  •  PubChem Substance ID 329752831

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Building Blocks, C6 to C8, Chemical Synthesis,
grade   for AOX determination (according to DIN 38409-H14)
InChI Key   WXNZTHHGJRFXKQ-UHFFFAOYSA-N
concentration   725 mg/L in H2O
bp   100 °C
density   1.00 g/mL at 20 °C

Description

Testing Method

ISO 9562:2004

Packaging

50 mL in glass bottle

Application

4-chlorophenol was used as a reagent, during development of high analytical performance-tyrosinase biosensor.

General description

4-chlorophenol dechlorinates to phenol rapidly, on palladized graphite electrodes and palladized carbon cloth.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?