EMAIL THIS PAGE TO A FRIEND
25895 Sigma-Aldrich

Clofibrate

≥98.0% (GC)

Synonym: 2-(4-Chlorophenoxy)-2-methylpropionic acid ethyl ester, Ethyl 2-(4-chlorophenoxy)-2-methylpropionate, Ethyl 2-(4-chlorophenoxy)isobutyrate

  • CAS Number 637-07-0

  • Empirical Formula (Hill Notation) C12H15ClO3

  • Molecular Weight 242.70

  •  Beilstein Registry Number 1913459

  •  EC Number 211-277-4

  •  MDL number MFCD00000615

Purchase

Properties

Related Categories Building Blocks, C12 to C63, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   KNHUKKLJHYUCFP-UHFFFAOYSA-N
assay   ≥98.0% (GC)
refractive index   n20/D 1.503
density   1.137 g/mL at 20 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3
RTECS 
UE9480000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?