EMAIL THIS PAGE TO A FRIEND
26388 Sigma-Aldrich

Chlorosulfonic acid

Purchase

Properties

Related Categories Acids, Acids & Bases, Analytical Reagents, Analytical/Chromatography, Chemical Synthesis,
Quality Level   PREMIUM
vapor density   4 (vs air)
vapor pressure   1 mmHg ( 25 °C)
  3.3 mmHg ( 37.7 °C)
InChI Key   XTHPWXDJESJLNJ-UHFFFAOYSA-N
expl. lim.   37.7 %
refractive index   n20/D 1.433(lit.)
bp   151-152 °C/755 mmHg(lit.)
density   1.753 g/mL at 25 °C(lit.)

Description

Caution

may contain particles and discolor to slightly greenish-yellow on storage

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 1754 8 / PGI
WGK Germany 
1
RTECS 
FX5730000
Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?