• USA Home
  • 26470 - O-Phenyl chlorothionoformate

EMAIL THIS PAGE TO A FRIEND
26470 Sigma-Aldrich

O-Phenyl chlorothionoformate

purum, ≥99.0% (AT)

Synonym: Phenyl thionochloroformate

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thiocarbonyl Compounds More...
InChI Key   KOSYAAIZOGNATQ-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (AT)
refractive index   n20/D 1.581(lit.)
  n20/D 1.581
bp   81-83 °C/6 mmHg(lit.)
density   1.248 g/mL at 25 °C(lit.)

Description

Other Notes

Reagent used for the Bu3SnH mediated deoxigenation of alcohols via thionocarbonates

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3265 8 / PGII
WGK Germany 
3
Flash Point(F) 
177.8 °F
Flash Point(C) 
81 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

234524 O-Phenyl chlorothionoformate, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?