EMAIL THIS PAGE TO A FRIEND
26500 Sigma-Aldrich

2-Chlorothiophene

technical, ≥95.0% (GC)

Synonym: 2-Thienyl chloride

  • CAS Number 96-43-5

  • Empirical Formula (Hill Notation) C4H3ClS

  • Molecular Weight 118.58

  •  Beilstein Registry Number 104652

  •  EC Number 202-505-3

  •  MDL number MFCD00005421

  •  PubChem Substance ID 329752861

Purchase

Properties

InChI Key   GSFNQBFZFXUTBN-UHFFFAOYSA-N
grade   technical
assay   ≥95.0% (GC)
refractive index   n20/D 1.547(lit.)
  n20/D 1.547
bp   127-129 °C(lit.)
density   1.286 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
73.4 °F
Flash Point(C) 
23 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

155667 2-Chlorothiophene, 96%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?