EMAIL THIS PAGE TO A FRIEND
26550 Sigma-Aldrich

4-Chlorotoluene

purum, ≥99.0% (GC)

Purchase

Properties

Related Categories Aryl, Building Blocks, C7, Chemical Synthesis, Halogenated Hydrocarbons,
vapor density   4.38 (vs air)
vapor pressure   10 mmHg ( 45 °C)
InChI Key   NPDACUSDTOMAMK-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
refractive index   n20/D 1.52(lit.)
  n20/D 1.521
bp   162 °C(lit.)
mp   6-8 °C(lit.)
density   1.07 g/mL at 25 °C(lit.)

Description

Other Notes

This product has been replaced by 24254-Sigma-Aldrich | Trichloroethylene, ≥99% | ClCH=CCl2

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2238 3 / PGIII
WGK Germany 
2
RTECS 
XS9010000
Flash Point(F) 
127.4 °F
Flash Point(C) 
53 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?