• USA Home
  • 26590 - 5-Chlorovaleronitrile

EMAIL THIS PAGE TO A FRIEND
26590 Sigma-Aldrich

5-Chlorovaleronitrile

purum, ≥95.0% (GC)

Synonym: 5-Chloropentanenitrile

  • CAS Number 6280-87-1

  • Linear Formula Cl(CH2)4CN

  • Molecular Weight 117.58

  •  Beilstein Registry Number 1742079

  •  EC Number 228-481-4

  •  MDL number MFCD00001977

Purchase

Properties

InChI Key   JSAWFGSXRPCFSW-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
refractive index   n20/D 1.446(lit.)
  n20/D 1.447
density   1.045 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3276 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
208.4 °F
Flash Point(C) 
98 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

C73001 5-Chlorovaleronitrile, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?