EMAIL THIS PAGE TO A FRIEND
26616 Sigma-Aldrich

Hydrogen chloride

puriss., ≥99.7%

  • CAS Number 7647-01-0

  • Linear Formula HCl

  • Molecular Weight 36.46

  •  Beilstein Registry Number 1098214

  •  MDL number MFCD00011324

  •  PubChem Substance ID 329752871

Purchase

Properties

Related Categories Chemical Synthesis, Specialty Gases, Synthetic Reagents More...
InChI Key   VEXZGXHMUGYJMC-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.7%

Description

Other Notes

Sales restrictions may apply

Packaging

Cylinder with net ~25 kg

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1050 - class 2 - Hydrogen chloride, anhydrous
WGK Germany 
1
RTECS 
MW4025000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?