EMAIL THIS PAGE TO A FRIEND
266345 Sigma-Aldrich

Zinc

granular, −10-+50 mesh, ≥99.8%, ACS reagent

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Materials Science, Metal and Ceramic Science, Metals,
grade   ACS reagent
vapor pressure   1 mmHg ( 487 °C)
InChI Key   HCHKCACWOHOZIP-UHFFFAOYSA-N
assay   ≥99.8%
form   granular
resistivity   5.8 μΩ-cm, 20°C
particle size   −10-+50 mesh
bp   907 °C(lit.)
mp   420 °C(lit.)
density   7.133 g/mL at 25 °C(lit.)
cation traces   Fe: ≤0.01%
  Pb: ≤0.01%

Description

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?