• USA Home
  • 27043 - Chondroitin 6-sulfate sodium salt from shark cartilage

EMAIL THIS PAGE TO A FRIEND
27043 Sigma-Aldrich

Chondroitin 6-sulfate sodium salt from shark cartilage

Synonym: Chondroitin sulfate C sodium salt, Poly[β-glucuronic acid-(1→3)-N-acetyl-β-galactosamine-6-sulfate-(1→4)] alternating

Purchase

Properties

Related Categories Carbohydrates, Cell Biology, Functional Foods, Glycosaminoglycan, Nutrition Research More...
optical activity   [α]20/D −15±5°, c = 1% in H2O
impurities   ~3% total nitrogen (N)
  ~7% total S
loss   ≤10% loss on drying
solubility   H2O: 10 mg/mL clear, colorless
suitability   suitable as substrate for chondroitinase test

Description

Application

Chondroitin, a glucosaminoglycan (GAG), is a polysaccharide chain of alternating units of N-acetylgalactosamine (GalNAc) and glucuronic acid (GlcA) that can be sulfated on the either or both GalNAc and GlcA units. Chondroitin and its sulfates are frequently attached to proteins to form proteoglycans.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C4384 Chondroitin sulfate sodium salt from shark cartilage, sulfated glycosaminoglycan polysaccharide

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?