270466 Sigma-Aldrich

2-Methyl-1-propanol

CHROMASOLV®, for HPLC, 99.5%

DOWNLOAD MSDS (PDF)

Synonym: Isobutanol, Isobutyl alcohol

Properties

Related Categories Amber Glass Bottles, Analytical Reagents, Analytical/Chromatography, CHROMASOLV for HPLC, Chromatography Reagents & Solvents,
grade   CHROMASOLV®
  for HPLC
vapor density   2.55 (vs air)
vapor pressure   8 mmHg ( 20 °C)
  8.8 mmHg ( 0 °C)
assay   99.5%
autoignition temp.   801 °F
purified by   glass distillation
expl. lim.   10.6 %
impurities   <0.050% water
evapn. residue   <0.0005%
refractive index   n20/D 1.396(lit.)
bp   108 °C(lit.)
mp   −108 °C(lit.)
density   0.803 g/mL at 25 °C(lit.)
λ   H2O reference
UV absorption   λ: 219 nm Amax: 1.00
  λ: 230 nm Amax: 0.50
  λ: 245 nm Amax: 0.10
  λ: 250 nm Amax: 0.05
  λ: 260 nm Amax: 0.03
  λ: 330-400 nm Amax: 0.01

Description

Packaging

1, 2 L in glass bottle

100 mL in glass bottle

Legal Information

CHROMASOLV is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH

Price and Availability

Customers Also Viewed

2-Methyl-1-propanol

BioUltra, for molecular biology, ≥99.5% (GC)

2-Methyl-1-propanol

analytical standard

2-Methyl-1-propanol

puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)

2-Methyl-1-propanol

anhydrous, 99.5%

Isobutyl alcohol

≥99%, FCC, FG

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?