Synonym: DMF
CAS Number 68-12-2
Linear Formula HCON(CH3)2
Molecular Weight 73.09
Beilstein Registry Number 605365
EC Number 200-679-5
MDL number MFCD00003284
PubChem Substance ID 24856379
Popular Documents: Product Information Sheet (PDF) | Specification Sheet (PDF)
| Related Categories | Amber Glass Bottles, Analytical Reagents, Analytical/Chromatography, CHROMASOLV Plus, Chromatography Reagents & Solvents, |
| grade | CHROMASOLV® Plus |
| for HPLC | |
| vapor density | 2.5 (vs air) |
| vapor pressure | 2.7 mmHg ( 20 °C) |
| assay | ≥99.9% |
| autoignition temp. | 833 °F |
| purified by | glass distillation |
Solvent for many hydrophobic organic compounds.
1, 6×1, 2, 4×2, 4×4 L in glass bottle
10 L in ss drum
100 mL in glass bottle
200 L in Pure-Pac™ 1
CHROMASOLV is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?