CAS Number 75-15-0
Empirical Formula (Hill Notation) CS2
Molecular Weight 76.14
Beilstein Registry Number 1098293
EC Number 200-843-6
MDL number MFCD00011321
PubChem Substance ID 24856387
Popular Documents: Specification Sheet (PDF)
| Related Categories | Analytical Reagents, Analytical/Chromatography, Chromatography Reagents & Solvents, HPLC & UHPLC Solvents (CHROMASOLV), HPLC Grade Solvents (CHROMASOLV), |
| grade | CHROMASOLV® |
| for HPLC | |
| vapor density | 2.67 (vs air) |
| vapor pressure | 19.48 psi ( 55 °C) |
| assay | ≥99.9% |
| autoignition temp. | 212 °F |
| expl. lim. | 50 % |
1, 2 L in Sure/Seal™
100 mL in Sure/Seal™
CHROMASOLV is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?