270709 Sigma-Aldrich

Benzene

CHROMASOLV® Plus, for HPLC, ≥99.9%

DOWNLOAD MSDS (PDF)

Properties

Related Categories Amber Glass Bottles, Analytical Reagents, Analytical/Chromatography, Benzene and Benzene Solutions, CHROMASOLV Plus,
grade   CHROMASOLV® Plus
  for HPLC
vapor density   2.77 (vs air)
vapor pressure   166 mmHg ( 37.7 °C)
  74.6 mmHg ( 20 °C)
assay   ≥99.9%
autoignition temp.   1043 °F
expl. lim.   8 %
impurities   ≤0.02% water (Karl Fischer)
evapn. residue   <0.0005%
color   APHA: ≤20
refractive index   n20/D 1.501(lit.)
bp   80 °C(lit.)
mp   5.5 °C(lit.)
density   0.874 g/mL at 25 °C(lit.)
λ   H2O reference
UV absorption   λ: 280 nm Amax: 1.0
  λ: 290 nm Amax: 0.15
  λ: 300 nm Amax: 0.06
  λ: 330 nm Amax: 0.02
  λ: 350-400 nm Amax: 0.01

Description

Biochem/physiol Actions

Environmental carcinogen; hematoxin that is linked to increased incidence of leukemia in humans.

Packaging

1, 6×1, 2, 4×4 L in glass bottle

100 mL in glass bottle

Legal Information

CHROMASOLV is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH

Price and Availability

Customers Also Viewed

Benzene
12540Benzene

analytical standard

Benzene
401765Benzene

anhydrous, 99.8%

Benzene
32212Benzene

puriss. p.a., Reag. Ph. Eur., ≥99.7% (GC)

Benzene
154628Benzene

ACS spectrophotometric grade, ≥99%

Benzene
319953Benzene

ACS reagent, ≥99.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?