270725 Sigma-Aldrich

Acetone

CHROMASOLV®, for HPLC, ≥99.9%

DOWNLOAD MSDS (PDF)

Properties

Related Categories Acetone, Amber Glass Bottles, Analytical Reagents, Analytical/Chromatography, CHROMASOLV for HPLC,
grade   CHROMASOLV®
  for HPLC
vapor density   2 (vs air)
vapor pressure   184 mmHg ( 20 °C)
assay   ≥99.9%
purified by   glass distillation
expl. lim.   13.2 %
impurities   ≤0.5% water
evapn. residue   ≤0.0002%
refractive index   n20/D 1.359(lit.)
bp   56 °C/760 mmHg(lit.)
mp   −94 °C(lit.)
density   0.791 g/mL at 25 °C(lit.)
UV absorption   λ: 330 nm Amax: 1.00
  λ: 340 nm Amax: 0.10
  λ: 350 nm Amax: 0.02
  λ: 400 nm Amax: 0.01

Description

Other Notes

Pure-Pac® II containers require the Micromatic MacroValve coupler for dispensing solvents, Z560723.

Packaging

1, 6×1, 2, 4×2, 4, 4×4 L in glass bottle

20, 50 L in Pure-Pac™ 2

View returnable container options.

100 mL in glass bottle

56, 200 L in Pure-Pac™ 1

Application

Acetone′s luminesence intensity is dependent upon the solution components 1. The absorption of UV light by acetone, results in its photolysis and the production of radials 1.

Legal Information

CHROMASOLV is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH

Pure-Pac is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Customers Also Viewed

Acetone
650501Acetone

CHROMASOLV® Plus, for HPLC, ≥99.9%

Acetone
34850Acetone

CHROMASOLV®, for HPLC, ≥99.8%

Acetone
439126Acetone

CHROMASOLV®, for HPLC, ≥99.9%

Acetone
414689Acetone

capillary GC grade, ≥99.9%

Acetone
179124Acetone

ACS reagent, ≥99.5%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?