EMAIL THIS PAGE TO A FRIEND

270733 Sigma-Aldrich

Water

CHROMASOLV®, for HPLC

Purchase

Properties

Related Categories Amber Glass Bottles, Analytical Reagents, Analytical/Chromatography, CHROMASOLV for HPLC, Chromatography Reagents & Solvents,
grade   CHROMASOLV®
  for HPLC
evapn. residue   <0.0003%
refractive index   n20/D 1.34(lit.)
bp   100 °C(lit.)
density   1.000 g/mL at 3.98 °C(lit.)
λ   H2O reference
UV absorption   λ: 205 nm Amax: 0.01
  λ: 210 nm Amax: 0.01
  λ: 250-400 nm Amax: 0.005

Description

Packaging

1, 6×1, 2.5, 4×2.5, 4, 4×4 L in glass bottle

18 L in VerSA-Flow™

20 L in Nowpak™

Preparation Note

Product filtered through a 0.2 μm filter

Legal Information

CHROMASOLV is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

Water
14263Water

LC-MS Ultra CHROMASOLV®, tested for UHPLC-MS

Water
39253Water

LC-MS CHROMASOLV®

Water
34877Water

CHROMASOLV® Plus, for HPLC

Water
95280Water

tested according to Ph.Eur.

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
nwg
RTECS 
ZC0110000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

Agriculture | Crop Protection Workflow

Pesticides are an essential component in modern agriculture and enable growers to fully realize the yield potential of their seeds. From compound discovery to active ingredient development, our chemi...
Keywords: Agriculture, Building blocks, Organic synthesis, Pesticides

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Water in Photosystem II: structural, functional and mechanistic considerations. Linke K and Ho FM Biochim. Biophys. Acta 1837(1), 14-32, (2014)

Loading...


Widespread decline of Congo rainforest greenness in the past decade. Zhou L, Tian Y, Myneni RB, et al. Nature 509(7498), 86-90, (2014)

Loading...


An antifreeze protein folds with an interior network of more than 400 semi-clathrate waters. Sun T, Lin FH, Campbell RL, et al. Science 343(6172), 795-8, (2014)

Loading...


Hydrophilic interaction chromatography versus reversed phase liquid chromatography coupled to mass spectrometry: effect of electrospray ionization source geometry on sensitivity. Periat A, Kohler I, Bugey A, et al. J. Chromatogr. A 1356, 211-20, (2014)

Loading...


PAH toxicity at aqueous solubility in the fish embryo test with Danio rerio using passive dosing. Seiler TB, Best N, Fernqvist MM, et al. Chemosphere 112, 77-84, (2014)

Loading...


Contrasting patterns of allochthony among three major groups of crustacean zooplankton in boreal and temperate lakes. Berggren M, Ziegler SE, St-Gelais NF, et al. Ecology 95(7), 1947-59, (2014)

Loading...


Tissue electrolytes in dehydrated children. Nutr. Rev. 25(7), 205-7, (1967)

Loading...


Cell volume, pumps, and neurologic function: brain's adaptation to osmotic stress. Fishman RA Res. Publ. Assoc. Res. Nerv. Ment. Dis. 53, 159-71, (1974)

Loading...


Ecological feedbacks. Termite mounds can increase the robustness of dryland ecosystems to climatic change. Bonachela JA, Pringle RM, Sheffer E, et al. Science 347(6222), 651-5, (2015)

Loading...


Present status of protozoan pathogens causing water-borne disease in northern part of El-Minia Governorate, Egypt. Khalifa RM, Ahmad AK, Abdel-Hafeez EH, et al. J. Egypt. Soc. Parasitol. 44(3), 559-66, (2014)

Loading...


Agriculture: State-of-the-art soil. Cernansky R Nature 517(7534), 258-60, (2015)

Loading...


A new wrinkle. Burgdorf WH and Hoenig LJ JAMA Dermatol. 151(1), 14, (2015)

Loading...


Thermal conductivity characteristics of dewatered sewage sludge by thermal hydrolysis reaction. Song HW, Park KJ, Han SK, et al. J. Air Waste Manag. Assoc. 64(12), 1384-9, (2014)

Loading...


Whole-body PET/CT evaluation of tumor perfusion using generator-based 62Cu-ethylglyoxal bis(thiosemicarbazonato)copper(II): validation by direct comparison to 15O-water in metastatic renal cell carcinoma. Fletcher JW, Logan TF, Eitel JA, et al. J. Nucl. Med. 56(1), 56-62, (2015)

Loading...


Microtensile bond strength of lithium disilicate ceramics to resin adhesives. Aboushelib MN and Sleem D J. Adhes. Dent. 16(6), 547-52, (2014)

Loading...


Self-adhesive resin cements: adhesive performance to indirect restorative ceramics. Zorzin J, Belli R, Wagner A, et al. J. Adhes. Dent. 16(6), 541-6, (2014)

Loading...


Bond strength and monomer conversion of indirect composite resin restorations, Part 1: Light vs heat polymerization. Malta DA, Magne P, and Monteiro-Junior S J. Adhes. Dent. 16(6), 517-22, (2014)

Loading...


Bond strength of novel CAD/CAM restorative materials to self-adhesive resin cement: the effect of surface treatments. Elsaka SE J. Adhes. Dent. 16(6), 531-40, (2014)

Loading...


Durability of fiber post-to-composite bonds achieved by physical vapor deposition and tribochemical silica coating. Rathke A, Frehse H, Muche R, et al. J. Adhes. Dent. 16(6), 559-65, (2014)

Loading...


Mapping the gating and permeation pathways in the voltage-gated proton channel Hv1. Chamberlin A, Qiu F, Wang Y, et al. J. Mol. Biol. 427(1), 131-45, (2015)

Loading...


Regulation of auxin, abscisic acid and salicylic acid levels by ascorbate application under heat stress in sensitive and tolerant maize leaves. Dinler BS, Demir E, and Kompe YO Acta Biol. Hung. 65(4), 469-80, (2014)

Loading...


Water used to moisten vegetables is a source of Escherichia coli and protozoan parasite contamination at markets in Hanoi, Vietnam. Tram NT and Dalsgaard A J. Water Health 12(4), 896-900, (2014)

Loading...


Optimization of factors affecting hexavalent chromium removal from simulated electroplating wastewater by synthesized magnetite nanoparticles. Ataabadi M, Hoodaji M, Tahmourespour A, et al. Environ. Monit. Assess. 187(1), 4165, (2015)

Loading...


Native structure of photosystem II at 1.95 Å resolution viewed by femtosecond X-ray pulses. Suga M, Akita F, Hirata K, et al. Nature 517(7532), 99-103, (2015)

Loading...


Reply: To PMID 23954778. Andreetti C, D'Andrilli A, Menna C, et al. Ann. Thorac. Surg. 98(6), 2274, (2014)

Loading...


Thoracoscopic water pleurectomy in pneumothorax: some concerns. Rena O, Papalia E, Turello D, et al. Ann. Thorac. Surg. 98(6), 2273-4, (2014)

Loading...


Density, ultrasound velocity, acoustic impedance, reflection and absorption coefficient determination of liquids via multiple reflection method. Hoche S, Hussein MA, and Becker T Ultrasonics 57, 65-71, (2015)

Loading...


Toxicity of Bacillus thuringiensis var. israelensis in aqueous suspension on the South American common frog Leptodactylus latrans (Anura: Leptodactylidae) tadpoles. Lajmanovich RC, Junges CM, Cabagna-Zenklusen MC, et al. Environ. Res. 136, 205-12, (2015)

Loading...


Relationship between ecophysiological factors, growth and ochratoxin A contamination of dry-cured sausage based matrices. Rodríguez A, Capela D, Medina Á, et al. Int. J. Food Microbiol. 194, 71-7, (2015)

Loading...


In vitro and in vivo evaluation of water-soluble iminophosphorane ruthenium(II) compounds. A potential chemotherapeutic agent for triple negative breast cancer. Frik M, Martínez A, Elie BT, et al. J. Med. Chem. 57(23), 9995-10012, (2014)

Loading...


Modeling the radiolysis of supercritical water by fast neutrons: density dependence of the yields of primary species at 400°c. Butarbutar SL, Meesungnoen J, Guzonas DA, et al. Radiat. Res. 182(6), 695-704, (2014)

Loading...


Crystal structure of a DNA/Ba2+ G-quadruplex containing a water-mediated C-tetrad. Zhang D, Huang T, Lukeman PS, et al. Nucleic Acids Res. 42(21), 13422-9, (2014)

Loading...


Distribution volumes of macromolecules in human ovarian and endometrial cancers--effects of extracellular matrix structure. Haslene-Hox H, Oveland E, Woie K, et al. Am. J. Physiol. Heart Circ. Physiol. 308(1), H18-28, (2015)

Loading...


Age-related changes in rat dermal extracellular matrix composition affect the distribution of plasma proteins as a function of size and charge. Sagstad SJ, Oveland E, Karlsen TV, et al. Am. J. Physiol. Heart Circ. Physiol. 308(1), H29-38, (2015)

Loading...


Evaluating cold, wind, and moisture protection of different coverings for prehospital maritime transportation-a thermal manikin and human study. Jussila K, Rissanen S, Parkkola K, et al. Prehosp. Disaster Med. 29(6), 580-8, (2014)

Loading...


The contribution of unimproved water and toilet facilities to pregnancy-related mortality in Afghanistan: analysis of the Afghan Mortality Survey. Gon G, Monzon-Llamas L, Benova L, et al. Trop. Med. Int. Health 19(12), 1488-99, (2014)

Loading...


Florfenicol residues in rainbow trout after oral dosing in recirculating and flow-through culture systems. Meinertz JR, Hess KR, Bernardy JA, et al. J. Aquat. Anim. Health 26(4), 243-50, (2014)

Loading...


Modelling of the growth/no growth interface of Wallemia sebi and Eurotium herbariorum as a function of pH, aw and ethanol concentration. Deschuyffeleer N, Vermeulen A, Daelman J, et al. Int. J. Food Microbiol. 192, 77-85, (2015)

Loading...


Comparative profiling of membrane lipids during water stress in Thellungiella salsuginea and its relative Arabidopsis thaliana. Yu B and Li W Phytochemistry 108, 77-86, (2014)

Loading...


Growth/no growth models for Zygosaccharomyces rouxii associated with acidic, sweet intermediate moisture food products. Marvig CL, Kristiansen RM, and Nielsen DS Int. J. Food Microbiol. 192, 51-7, (2015)

Loading...


Simultaneous exposure to Escherichia coli heat-labile and heat-stable enterotoxins increases fluid secretion and alters cyclic nucleotide and cytokine production by intestinal epithelial cells. Read LT, Hahn RW, Thompson CC, et al. Infect. Immun. 82(12), 5308-16, (2014)

Loading...


Does water-jet force make a difference in fat grafting? In vitro and in vivo evidence of improved lipoaspirate viability and fat graft survival. Yin S, Luan J, Fu S, et al. Plast. Reconstr. Surg. 135(1), 127-38, (2015)

Loading...


Microleakage of inlay ceramic systems luted with self-adhesive resin cements. Uludag B, Yucedag E, and Sahin V J. Adhes. Dent. 16(6), 523-9, (2014)

Loading...


Office construction or remodeling: are you going to LEED in 2014? Knowles L J. Mich. Dent. Assoc. 96(7), 34-5, (2014)

Loading...


Temperance and Genevieve. Mingus L Midwifery Today Int. Midwife (110), 43-4, (2014)

Loading...


Rose and Philippa. Whistler J Midwifery Today Int. Midwife (110), 42-3, (2014)

Loading...


Jade and Cole. Bernholz SL Midwifery Today Int. Midwife (110), 39-41, (2014)

Loading...


Response of maize serine/arginine-rich protein gene family in seedlings to drought stress. Li J, Guo Y, Cui W, et al. Yi Chuan 36(7), 697-706, (2014)

Loading...


ACOG Committee Opinion no. 594: Immersion in water during labor and delivery. Obstet. Gynecol. 123(4), 912-5, (2014)

Loading...


Comparison of fracture resistance and fracture characterization of bilayered zirconia/fluorapatite and monolithic lithium disilicate all ceramic crowns. Altamimi AM, Tripodakis AP, Eliades G, et al. Int. J. Esthet. Dent. 9(1), 98-110, (2014)

Loading...


Micromorphology of ceramic etching pattern for two CAD-CAM and one conventional feldspathic porcelain and need for post-etching cleaning. Onisor I, Rocca GT, and Krejci I Int. J. Esthet. Dent. 9(1), 54-69, (2014)

Loading...


Heat removal using microclimate foot cooling: a thermal foot manikin study. Castellani JW, Demes R, Endrusick TL, et al. Aviat. Space. Environ. Med. 85(4), 445-8, (2014)

Loading...


Helicopter crashes into water: warning time, final position, and other factors affecting survival. Brooks CJ, MacDonald CV, Baker SP, et al. Aviat. Space. Environ. Med. 85(4), 440-4, (2014)

Loading...


Evaluation of the effect of processing methods on the nutritional and anti-nutritional compositions of two under-utilized Nigerian grain legumes. Oke MO, Sobowale SS, and Ogunlakin GO Pak. J. Biol. Sci. 16(24), 2015-20, (2013)

Loading...


Influence of water stress on antioxidative enzymes and yield of banana cultivars and hybrids. Surendar KK, Devi DD, Ravi I, et al. Pak. J. Biol. Sci. 16(24), 1997-2002, (2013)

Loading...


Behavioral pattern of physicochemical constituents of the postharvest mango (Mangifera indica L.) influenced by storage stimuli. Islam MK Pak. J. Biol. Sci. 16(24), 1985-90, (2013)

Loading...


Proximate and phytochemical of Cola nitida and Cola acuminata. Dewole EA, Dewumi DF, Alabi JY, et al. Pak. J. Biol. Sci. 16(22), 1593-6, (2013)

Loading...


Synthesis of carboxymethyl chitosan and coating on wound dressing gauze for wound healing. Venkatrajah B, Malathy VV, Elayarajah B, et al. Pak. J. Biol. Sci. 16(22), 1438-48, (2013)

Loading...


Temperature and saline stress on seedlings of Swietenia macrophylla: a comparative study. Rahman MS, Akter S, and Al-Amin M Pak. J. Biol. Sci. 16(23), 1765-70, (2013)

Loading...


[Light-isotope water as a therapeutic agent for acute radiation disease]. Ivanov AA, Ushakov IB, Kulikova EI, et al. Aviakosm. Ekolog. Med. 47(5), 40-4, (2013)

Loading...


[Modified HPLC method of determination of the valproic acid in biological fluids]. Dutov AA, Nikitin DA, Letunov VI, et al. Biomed. Khim. 59(5), 514-8, (2013)

Loading...


[Identification of components of the milky juice of Lactarius pergamenus (Fr.)Fr fungi by gas-liquid chromatography/mass spectrometry]. Tsyvins'ka MV, Panchak LV, Stoĭka RS, et al. Ukr. Biokhim. Zh. 85(5), 170-6, (2013)

Loading...


[NAC transcription factors family and increased tolerance to water deficiency in plants]. Lip S, Kurowska M, and Szarejko I Postepy Biochem. 59(3), 295-304, (2013)

Loading...


[The cutting-edge of medicine; hyponatremia and water metabolism disorders in elderly]. Ishikawa SE Nippon. Naika Gakkai Zasshi. 102(7), 1807-13, (2013)

Loading...


[Basic characteristics and clinical application of ozone nano-bubble water]. Arakawa S Kokubyo. Gakkai Zasshi. 80(1), 1-7, (2013)

Loading...


A two-compartment photoelectrochemical cell for solar hydrogen production. Rongé J and Martens JA Commun. Agric. Appl. Biol. Sci. 78(1), 179-84, (2013)

Loading...


[Water for the preparation of upper gastrointestinal endoscopy after gastrectomy]. Mukaide H, Inoue K, Fukui J, et al. Gan To Kagaku Ryoho. 40(5), 609-12, (2013)

Loading...


Quantitative cw Overhauser effect dynamic nuclear polarization for the analysis of local water dynamics. Franck JM, Pavlova A, Scott JA, et al. Prog. Nucl. Magn. Reson. Spectrosc. 74, 33-56, (2013)

Loading...


Carbon stocks of intact mangroves and carbon emissions arising from their conversion in the Dominican Republic. Kauffman JB, Heider C, Norfolk J, et al. Ecol. Appl. 24(3), 518-27, (2014)

Loading...


Photosynthesis. Electronic structure of the oxygen-evolving complex in photosystem II prior to O-O bond formation. Cox N, Retegan M, Neese F, et al. Science 345(6198), 804-8, (2014)

Loading...


The role of protein-bound water molecules in microbial rhodopsins. Gerwert K, Freier E, and Wolf S Biochim. Biophys. Acta 1837(5), 606-13, (2014)

Loading...


Freeze-fracture shadow-casting (FreSCA) cryo-SEM as a tool to investigate the wetting of micro- and nanoparticles at liquid-liquid interfaces. Isa L Chimia 67(4), 231-5, (2013)

Loading...


The governance dimensions of water security: a review. Bakker K and Morinville C Philos. Transact. A. Math. Phys. Eng. Sci. 371(2002), 20130116, (2013)

Loading...


Toward a mechanism for eastern North American forest mesophication: differential litter drying across 17 species. Kreye JK, Varner JM, Hiers JK, et al. Ecol. Appl. 23(8), 1976-86, (2013)

Loading...


Effects of thinning on drought vulnerability and climate response in north temperate forest ecosystems. D'Amato AW, Bradford JB, Fraver S, et al. Ecol. Appl. 23(8), 1735-42, (2013)

Loading...


Direct bacterial profiling by matrix-assisted laser desorption-ionization time-of-flight mass spectrometry for identification of pathogenic Neisseria. Elena N. Ilina et al J. Mol. Diagn. 11, 75-86, (2009)

Loading...


Future species composition will affect forest water use after loss of eastern hemlock from southern Appalachian forests. Brantley S, Ford CR, and Vose JM Ecol. Appl. 23(4), 777-90, (2013)

Loading...


Hydrogen-bonding changes of internal water molecules upon the actions of microbial rhodopsins studied by FTIR spectroscopy. Furutani Y and Kandori H Biochim. Biophys. Acta 1837(5), 598-605, (2014)

Loading...


OSCA1 mediates osmotic-stress-evoked Ca2+ increases vital for osmosensing in Arabidopsis. Yuan F, Yang H, Xue Y, et al. Nature 514(7522), 367-71, (2014)

Loading...


Lewis acid catalyzed cascade reaction to carbazoles and naphthalenes via dehydrative [3 + 3]-annulation. Wang S, Chai Z, Wei Y, et al. Org. Lett. 16(13), 3592-5, (2014)

Loading...


Proton transport facilitating water-oxidation: the role of second sphere ligands surrounding the catalytic metal cluster. Bao H, Dilbeck PL, and Burnap RL Photosynth. Res. 116(2-3), 215-29, (2013)

Loading...


A comparative study of Mn/CeO2, Mn/ZrO2 and Mn/Ce-ZrO2 for low temperature selective catalytic reduction of NO with NH3 in the presence of SO2 and H2O. Shen B, Zhang X, Ma H, et al. J. Environ. Sci. (China) 25(4), 791-800, (2013)

Loading...


Calculated and experimental chromatograms for distorted gradients and non-linear solvation strength retention models. Gritti F and Guiochon G J. Chromatogr. A 1356, 96-104, (2014)

Loading...


Evaluation of hydrolysis and alcoholysis reactions in gas chromatography/mass spectrometry inlets. Ai G, Sun T, and Dong X J. Chromatogr. A 1356, 283-8, (2014)

Loading...


Separations by gradient elution: why are steep gradient profiles distorted and what is their impact on resolution in reversed-phase liquid chromatography. Gritti F and Guiochon G J. Chromatogr. A 1344, 66-75, (2014)

Loading...


Effects on nano zero-valent iron reactivity of interactions between hardness, alkalinity, and natural organic matter in reverse osmosis concentrate. Hwang Y and Shin HS J. Environ. Sci. (China) 25(11), 2177-84, (2013)

Loading...


Separate and combined responses to water deficit and UV-B radiation. Bandurska H, Niedziela J, and Chadzinikolau T Plant Sci. 213, 98-105, (2013)

Loading...


Water-based solutions are the best decontaminating fluids for dermal corrosive exposures: a mini review. Brent J Clin. Toxicol. (Phila.) 51(8), 731-6, (2013)

Loading...


Recent advances in transport of water-soluble vitamins in organs of the digestive system: a focus on the colon and the pancreas. Said HM Am. J. Physiol. Gastrointest. Liver Physiol. 305(9), G601-10, (2013)

Loading...


Electrochemical detection and degradation of ibuprofen from water on multi-walled carbon nanotubes-epoxy composite electrode. Motoc S, Remes A, Pop A, et al. J. Environ. Sci. (China) 25(4), 838-47, (2013)

Loading...


Calcium-dependent behavioural responses to acute copper exposure in Oncorhynchus mykiss. Poulsen SB, Svendsen JC, Aarestrup K, et al. J. Fish Biol. 84(5), 1326-39, (2014)

Loading...


Assessing the impact of a school-based latrine cleaning and handwashing program on pupil absence in Nyanza Province, Kenya: a cluster-randomized trial. Caruso BA, Freeman MC, Garn JV, et al. Trop. Med. Int. Health 19(10), 1185-97, (2014)

Loading...


Computational design of water-soluble α-helical barrels. Thomson AR, Wood CW, Burton AJ, et al. Science 346(6208), 485-8, (2014)

Loading...


Utilization of carbon nanotubes for the removal of rhodamine B dye from aqueous solutions. Kumar S, Bhanjana G, Jangra K, et al. J. Nanosci. Nanotechnol. 14(6), 4331-6, (2014)

Loading...


[Dental fluorosis prevalence in Mexican localities of 27 states and the D.F.: six years after the publication of the Salt Fluoridation Mexican Official Regulation]. Betancourt-Lineares A, Irigoyen-Camacho ME, Mejía-González A, et al. Rev. Invest. Clin. 65(3), 237-47, (2013)

Loading...


Ion permeation in K⁺ channels occurs by direct Coulomb knock-on. Köpfer DA, Song C, Gruene T, et al. Science 346(6207), 352-5, (2014)

Loading...


Development of artificial dermis using 3D electrospun silk fibroin nanofiber matrix. Lee OJ, Ju HW, Kim JH, et al. J. Biomed. Nanotechnol. 10(7), 1294-303, (2014)

Loading...


Removal of Pb(II) from aqueous solution by hydrous manganese dioxide: adsorption behavior and mechanism. Xu M, Wang H, Di L, et al. J. Environ. Sci. (China) 25(3), 479-86, (2013)

Loading...


An Earth-sized planet in the habitable zone of a cool star. Quintana EV, Barclay T, Raymond SN, et al. Science 344(6181), 277-80, (2014)

Loading...


Pressure-induced gelatinization of starch in excess water. Vallons KJ, Ryan LA, and Arendt EK Crit. Rev. Food Sci. Nutr 54(3), 399-409, (2014)

Loading...


Pupal development of Aethina tumida (Coleoptera: Nitidulidae) in thermo-hygrometric soil conditions encountered in temperate climates. Bernier M, Fournier V, and Giovenazzo P J. Econ. Entomol. 107(2), 531-7, (2014)

Loading...


Recent progress in the applications of layer-by-layer assembly to the preparation of nanostructured ion-rejecting water purification membranes. Sanyal O and Lee I J. Nanosci. Nanotechnol. 14(3), 2178-89, (2014)

Loading...


Centennial-scale changes in the global carbon cycle during the last deglaciation. Marcott SA, Bauska TK, Buizert C, et al. Nature 514(7524), 616-9, (2014)

Loading...


Water channel structures analysed by electron crystallography. Tani K and Fujiyoshi Y Biochim. Biophys. Acta 1840(5), 1605-13, (2014)

Loading...


Porous coordination polymers as novel sorption materials for heat transformation processes. Janiak C and Henninger SK Chimia 67(6), 419-24, (2013)

Loading...


Water condensation: a multiscale phenomenon. Jensen KR, Fojan P, Jensen RL, et al. J. Nanosci. Nanotechnol. 14(2), 1859-71, (2014)

Loading...


Seasonal variation and source apportionment of organic and inorganic compounds in PM2.5 and PM10 particulates in Beijing, China. Li X, Wang Y, Guo X, et al. J. Environ. Sci. (China) 25(4), 741-50, (2013)

Loading...


Flow field and dissolved oxygen distributions in the outer channel of the Orbal oxidation ditch by monitor and CFD simulation. Guo X, Zhou X, Chen Q, et al. J. Environ. Sci. (China) 25(4), 645-51, (2013)

Loading...


Effects of solution chemistry on arsenic(V) removal by low-cost adsorbents. Wang Y and Tsang DC J. Environ. Sci. (China) 25(11), 2291-8, (2013)

Loading...


Swift adsorptive removal of Congo red from aqueous solution by K1.33Mn8O16 nanowires. Wu J, Li H, Wang J, et al. J. Nanosci. Nanotechnol. 13(8), 5452-60, (2013)

Loading...


Correction of water content and solute concentration in blood during hemoconcentration. Ohira Y, Ito A, and Ikawa S J. Appl. Physiol. 42(5), 739-43, (1977)

Loading...


Soluble microbial products from water biological treatment process: a review. Kang J, Du G, Gao X, et al. Water Environ. Res. 86(3), 223-31, (2014)

Loading...


Mercury bioaccumulation and bioaccumulation factors for Everglades mosquitofish as related to sulfate: a re-analysis of Julian II (2013). Pollman CD and Axelrad DM Bull. Environ. Contam. Toxicol. 93(5), 509-16, (2014)

Loading...


Ultrasound-assisted surfactant-enhanced emulsification microextraction for the determination of Cd and Ni in tea and water samples. Ezoddin M, Taghizadeh T, and Majidi B Environ. Technol. 35(17-20), 2401-9, (2014)

Loading...


Low-temperature selective catalytic reduction of NO on CeO2-CuO/Al2O3 catalysts prepared by different methods. Guo RT, Zhen WL, Pan WG, et al. Environ. Technol. 35(13-16), 1766-72, (2014)

Loading...


Effects of water-emulsified fuel on a diesel engine generator's thermal efficiency and exhaust. Syu JY, Chang YY, Tseng CH, et al. J. Air Waste Manag. Assoc. 64(8), 970-8, (2014)

Loading...


Characterization and humidity sensing properties of the sensor based on Na2Ti3O7 nanotubes. Zhang Y, Wu J, Zhang Y, et al. J. Nanosci. Nanotechnol. 14(6), 4303-7, (2014)

Loading...


Lewis acid-water/alcohol complexes as hydrogen atom donors in radical reactions. Povie G and Renaud P Chimia 67(4), 250-2, (2013)

Loading...


Methane emissions from rice fields under continuous straw return in the middle-lower reaches of the Yangtze River. Hou P, Li G, Wang S, et al. J. Environ. Sci. (China) 25(9), 1874-81, (2013)

Loading...


Capture of carbon dioxide from flue gas on TEPA-grafted metal-organic framework Mg2(dobdc). Cao Y, Song F, Zhao Y, et al. J. Environ. Sci. (China) 25(10), 2081-7, (2013)

Loading...


Distribution characteristics and source identification of polychlorinated dibenzo-p-dioxin and dibenzofurans, and dioxin-like polychlorinated biphenyls in the waters from River Kanzaki, running through Osaka urban area, Japan. Kishida M J. Environ. Sci. (China) 25(3), 441-51, (2013)

Loading...


Artificial receptors for the extraction of nucleoside metabolite 7-methylguanosine from aqueous media made by molecular imprinting. Krstulja A, De Schutter C, Favetta P, et al. J. Chromatogr. A 1365, 12-8, (2014)

Loading...


Adsorption of aqueous organic mixtures on a chiral stationary phase with bound antibiotic eremomycin. Nikitina YK, Ali I, and Asnin LD J. Chromatogr. A 1363, 71-8, (2014)

Loading...


Hydrophilic organic/salt-containing aqueous two-phase solvent system for counter-current chromatography: a novel technique for separation of polar compounds. Liu D, Zou X, Gao M, et al. J. Chromatogr. A 1356, 157-62, (2014)

Loading...


Optimizations of packed sorbent and inlet temperature for large volume-direct aqueous injection-gas chromatography to determine high boiling volatile organic compounds in water. Yu B, Song Y, Han L, et al. J. Chromatogr. A 1356, 221-9, (2014)

Loading...


Performance of different C18 columns in reversed-phase liquid chromatography with hydro-organic and micellar-organic mobile phases. Ruiz-Angel MJ, Pous-Torres S, Carda-Broch S, et al. J. Chromatogr. A 1344, 76-82, (2014)

Loading...


The impact of water and temperature interactions on lag phase, growth and potential ochratoxin A production by two new species, Aspergillus aculeatinus and A. sclerotiicarbonarius, on a green coffee-based medium. Akbar A and Magan N Int. J. Food Microbiol. 188, 116-21, (2014)

Loading...


Bacterial maximum non-inhibitory and minimum inhibitory concentrations of different water activity depressing solutes. Cebrián G, Arroyo C, Mañas P, et al. Int. J. Food Microbiol. 188, 67-74, (2014)

Loading...


Simultaneous preconcentration and determination of Ni and Pb in water samples by solid-phase extraction and flame Atomic absorption spectrometry. Ciftci H, Temuz MM, and Ciftci E J. AOAC Int. 96(4), 875-9, (2013)

Loading...


Potential use of calcareous mudstones in low hydraulic conductivity earthen barriers for environmental applications. Musso TB, Francisca FM, Musso TB, et al. Environ. Technol. 34(17-20), 2465-76, (2013)

Loading...


Study of penetration behavior of PCB-DNAPL in a sand layer by a column experiment. Okuda N, Shimizu T, Muratani M, et al. Chemosphere 114, 59-68, (2014)

Loading...


Effects of gas compositions on NOx reduction by selective non-catalytic reduction with ammonia in a simulated cement precalciner atmosphere. Fan W, Zhu T, Sun Y, et al. Chemosphere 113, 182-7, (2014)

Loading...


Disinfection of Bacillus spores with acidified nitrite. Szabo JG, Adcock NJ, and Rice EW Chemosphere 113, 171-4, (2014)

Loading...


Fate of phosphorus in diluted urine with tap water. Liu X, Wen G, Wang H, et al. Chemosphere 113, 146-50, (2014)

Loading...


The effects of acrylamide polyelectrolytes on aquatic organisms: relating toxicity to chain architecture. Costa R, Pereira JL, Gomes J, et al. Chemosphere 112, 177-84, (2014)

Loading...


Mechanical evaluation of four narrow-diameter implant systems. Hirata R, Bonfante EA, Machado LS, et al. Int. J. Prosthodont. 27(4), 359-62, (2014)

Loading...


Should water precautions be recommended for children with tympanostomy tubes? Wilcox LJ and Darrow DH Laryngoscope 124(1), 10-1, (2014)

Loading...


Nanofluids used for water/wastewater treatment--a mini review. Zhang L, Li Y, Liu X, et al. Recent Pat. Nanotechnol. 7(3), 224-31, (2013)

Loading...


Photodegradation of the antihistamine cetirizine in natural waters. Mead RN, Barefoot S, Helms JR, et al. Environ. Toxicol. Chem. 33(10), 2240-5, (2014)

Loading...


Water regime of Playa Lakes from southern Spain: conditioning factors and hydrological modeling. Moral F, Rodriguez-Rodriguez M, Beltrán M, et al. Water Environ. Res. 85(7), 632-42, (2013)

Loading...


Performance evaluation and modeling of synthetic-fiber barrier in the treatment of turbid water. Yu J and Kim Y Water Environ. Res. 85(7), 596-603, (2013)

Loading...


Metabolic engineering of cyanobacteria for the production of hydrogen from water. Rögner M Biochem. Soc. Trans. 41(5), 1254-9, (2013)

Loading...


Preparation and evaluation of Zr-beta-FeOOH for efficient arsenic removal. Sun X, Hu C, and Qu J J. Environ. Sci. (China) 25(4), 815-22, (2013)

Loading...


Effects of soil water and nitrogen availability on photosynthesis and water use efficiency of Robinia pseudoacacia seedlings. Liu X, Fan Y, Long J, et al. J. Environ. Sci. (China) 25(3), 585-95, (2013)

Loading...


Identification of causative compounds and microorganisms for musty odor occurrence in the Huangpu River, China. Sun D, Yu J, An W, et al. J. Environ. Sci. (China) 25(3), 460-5, (2013)

Loading...


Effects of sludge dredging on the prevention and control of algae-caused black bloom in Taihu Lake, China. He W, Shang J, Lu X, et al. J. Environ. Sci. (China) 25(3), 430-40, (2013)

Loading...


The behavioural response of adult Petromyzon marinus to damage-released alarm and predator cues. Imre I, Di Rocco RT, Belanger CF, et al. J. Fish Biol. 84(5), 1490-502, (2014)

Loading...


Positive symptoms and water diffusivity of the prefrontal and temporal cortices in schizophrenia patients: a pilot study. Park JY, Park HJ, Kim DJ, et al. Psychiatry Res. 224(1), 49-57, (2014)

Loading...


Impact of UV-B on drought- or cadmium-induced changes in the fatty acid composition of membrane lipid fractions in wheat. Gondor OK, Szalai G, Kovács V, et al. Ecotoxicol. Environ. Saf. 108, 129-34, (2014)

Loading...


Conductive poly(2,5-substituted aniline)s highly soluble both in water and organic solvents. Xu S, Ogi S, Sugiyasu K, et al. J. Nanosci. Nanotechnol. 14(6), 4449-54, (2014)

Loading...


Spatial and seasonal variability of CO2 flux at the air-water interface of the Three Gorges Reservoir. Le Y, Lu F, Wang X, et al. J. Environ. Sci. (China) 25(11), 2229-38, (2013)

Loading...


Mercury transportation in soil via using gypsum from flue gas desulfurization unit in coal-fired power plant. Wang K, Orndorff W, Cao Y, et al. J. Environ. Sci. (China) 25(9), 1858-64, (2013)

Loading...


Cr(VI) removal from aqueous solution with bamboo charcoal chemically modified by iron and cobalt with the assistance of microwave. Wang W, Wang X, Wang X, et al. J. Environ. Sci. (China) 25(9), 1726-35, (2013)

Loading...


Critical velocity in phosphorus exchange processes across the sediment-water interface. Wan J, Wang Z, Li Z, et al. J. Environ. Sci. (China) 25(10), 1966-71, (2013)

Loading...


Pesticide multiresidue determination in rice paddy water by gas chromatography coupled with triple quadrupole mass spectrometry. Pereira MB, Facco JF, Zemolin GM, et al. J. AOAC Int. 97(4), 987-94, (2014)

Loading...


Displacement-dispersive liquid-liquid microextraction based on solidification of floating organic drop of trace amounts of palladium in water and road dust samples prior to graphite furnace atomic absorption spectrometry determination. Ghanbarian M, Afzali D, Mostafavi A, et al. J. AOAC Int. 96(4), 880-6, (2013)

Loading...


Removal of phenolic compounds from aqueous phase by adsorption onto polymer supported iron nanoparticles. Sen BK, Deshmukh DK, Deb MK, et al. Bull. Environ. Contam. Toxicol. 93(5), 549-54, (2014)

Loading...


Synthesis of high-quality water-soluble near-infrared-emitting CdTe quantum dots capped with 3-mercaptobutyric acid. Ma KG, Bai JY, Fang T, et al. J. Nanosci. Nanotechnol. 14(7), 4940-8, (2014)

Loading...


Hydrothermal synthesis and photoluminescent properties of rod-shape assemblies of LaBO3:Eu3+ nanocrystals. Zeng Y, Tao Q, Li C, et al. J. Nanosci. Nanotechnol. 14(6), 4579-83, (2014)

Loading...


Refluxing synthesis, photoluminescence and binding ability to deoxyribonucleic acid of water-soluble rare earth ion-doped LaF3 nanoparticles. Wang Z, Zhang Y, Li C, et al. J. Nanosci. Nanotechnol. 14(6), 4506-12, (2014)

Loading...


Template-free hydrothermal synthesis different morphologies of visible-light-driven BiVO4 photocatalysts. Xu C, Zhu G, Wu J, et al. J. Nanosci. Nanotechnol. 14(6), 4475-80, (2014)

Loading...


Easily controllable synthesis of alpha-MoO3 nanobelts and MoO2 microaxletrees through one-pot hydrothermal route. Hu H, Xu J, Deng C, et al. J. Nanosci. Nanotechnol. 14(6), 4462-8, (2014)

Loading...


Water soluble sodium sulfate nanorods as a versatile template for the designing of copper sulfide nanotubes. Das G, Kakati N, Lee SH, et al. J. Nanosci. Nanotechnol. 14(6), 4455-61, (2014)

Loading...


Near-infrared to visible upconversion in Tm3+ and Yb3+ codoped Lu2O3 nanocrystals synthesized by hydrothermal method. Li L, Zhang X, Wei X, et al. J. Nanosci. Nanotechnol. 14(6), 4313-9, (2014)

Loading...


Effect of Cu2O morphology on photocatalytic hydrogen generation and chemical stability of TiO2/Cu2O composite. Zhu L, Zhang J, Chen Z, et al. J. Nanosci. Nanotechnol. 13(7), 5104-8, (2013)

Loading...


Dehydration of phenylboronic acid to boroxine catalyzed by Au(n) nanoclusters with atom packing core-shell structure. Huang P, Jiang Z, Chen G, et al. J. Nanosci. Nanotechnol. 13(7), 5088-92, (2013)

Loading...


Indoor water and dampness and the health effects on children: a review. Kennedy K and Grimes C Curr. Allergy Asthma Rep. 13(6), 672-80, (2013)

Loading...


Antibacterial efficacy and effect of chlorhexidine mixed with irreversible hydrocolloid for dental impressions: a randomized controlled trial. Cubas G, Valentini F, Camacho GB, et al. Int. J. Prosthodont. 27(4), 363-5, (2014)

Loading...


Analysis of the saturation phenomena of the neutralization rate of positively charged 218Po in water vapor. Tan Y, Xiao D, Shan J, et al. Health Phys. 107(3), 255-60, (2014)

Loading...


Analysis of fish DNA biomarkers as a molecular-level approach for ecological health assessments in an urban stream. Kang N, Kang HI, and An KG Bull. Environ. Contam. Toxicol. 93(5), 555-60, (2014)

Loading...


Analysis of cyazofamid and its metabolite in the environmental and crop samples using LC-MS/MS. Lee H, Kim E, Lee JH, et al. Bull. Environ. Contam. Toxicol. 93(5), 586-90, (2014)

Loading...


Cucumis sativus L-type lectin receptor kinase (CsLecRK) gene family response to Phytophthora melonis, Phytophthora capsici and water immersion in disease resistant and susceptible cucumber cultivars. Wu T, Wang R, Xu X, et al. Gene 549(2), 214-22, (2014)

Loading...


Cyclodextrin-based combinatorial polymers: efficient binders of pharmaceuticals in water. Shahgaldian P and Corvini PF Chimia 67(6), 425-6, (2013)

Loading...


Hydraulic conductivity of compacted soils controlled by microbial activity. Glatstein DA and Francisca FM Environ. Technol. 35(13-16), 1886-92, (2014)

Loading...


Degradation of triphenylborane-pyridine antifouling agent in water by copper ions. Tsuboi A, Okamura H, Kaewchuay N, et al. Environ. Technol. 34(17-20), 2835-40, (2013)

Loading...


Deposition and transport of Pseudomonas aeruginosa in porous media: lab-scale experiments and model analysis. Kwon KS, Kim SB, Choi NC, et al. Environ. Technol. 34(17-20), 2757-64, (2013)

Loading...


Magnetic alginate-layered double hydroxide composites for phosphate removal. Lee CG and Kim SB Environ. Technol. 34(17-20), 2749-56, (2013)

Loading...


Kinetics of degradation of imidapril hydrochloride in finished dosage formulations. Stanisz B and Regulska K Acta Pol. Pharm. 70(4), 737-42, (2013)

Loading...


In vitro anti-oxidative activities and GC-MS analysis of various solvent extracts of Cassia singueana parts. Ibrahim MA, Koorbanally NA, and Islam MS Acta Pol. Pharm. 70(4), 709-19, (2013)

Loading...


Endohedral complexes of fullerene C60 with small convalent molecules (H2O, NH3, H2, 2H2, 3H2, 4H2, O2, O3) in the context of potential drug transporter system. Medrek M, Pluciński F, and Mazurek AP Acta Pol. Pharm. 70(4), 659-65, (2013)

Loading...


Application of TLC method with video scanning in estimation of daily dietary intake of specific flavonoids--preliminary studies. Koch W, Kukuła-Koch W, Marzec Z, et al. Acta Pol. Pharm. 70(4), 611-20, (2013)

Loading...


Endothelium-dependent and independent vasorelaxant effects of aqueous extract of Tridax procumbens Lin. leaf in rat aortic rings. Salahdeen HM, Idowu GO, and Murtala BA Indian J. Exp. Biol. 50(12), 883-8, (2012)

Loading...


[Seedlings growth and survival of five Acacia (Fabaceae) species that coexists in neotropical semi-arid forests of Argentina, under different light and water availability conditions]. Venier P, Cabido M, Mangeaud A, et al. Rev. Biol. Trop. 61(2), 501-14, (2013)

Loading...


Effect of phenological stages on nutritional assessment of ten plant species in Tehsil Takht-e-Nasrati, District Karak, Pakistan. Khan M, Hussain F, Faridullah, et al. Pak. J. Pharm. Sci. 27(4), 953-61, (2014)

Loading...


New formula herbal pellets demonstrate a uniform and stable release of the active ingredients in vitro. Pranskuniene Z, Bernatoniene J, Kalveniene Z, et al. Acta Pol. Pharm. 70(4), 727-36, (2013)

Loading...


Effect of water soluble carrier on dissolution profiles of diclofenac sodium. Cwiertnia B Acta Pol. Pharm. 70(4), 721-6, (2013)

Loading...


Salt and water absorption by the intestinal tract. Parsons DS Br. Med. Bull. 23(3), 252-7, (1967)

Loading...


Material witness: watching wood dry. Ball P Nat. Mater. 13(10), 922, (2014)

Loading...


Aquagenic wrinkling: a unique facial presentation. Stamey C, Boos MD, Rosenbach M, et al. J. Am. Acad. Dermatol. 71(4), e150-2, (2014)

Loading...


Images in clinical medicine. Aquagenic keratoderma. Poletti ED and Muñoz-Sandoval R N. Engl. J. Med. 371(10), 952, (2014)

Loading...


Chemistry. New recipe produces ammonia from air, water, and sunlight. Service RF Science 345(6197), 610, (2014)

Loading...


Chronic pain localized to the iliohypogastric nerve: treatment using an ultrasound-guided technique of hydrodissection for catheter placement as a guide for surgical iliohypogastric nerve resection. Adler AC, Smith DI, and Parikh PM Plast. Reconstr. Surg. 134(1), 182e-183e, (2014)

Loading...


Astronomy. Almost-Earth tantalizes astronomers with promise of worlds to come. Bhattacharjee Y Science 344(6181), 249, (2014)

Loading...


Maritime biosecurity adrift. Hulme PE Science 343(6171), 611-2, (2014)

Loading...


IBI* series winner. An inquiry into the water around us. Saitta EK, Legron-Rodriguez T, and Bowdon MA Science 341(6149), 971-2, (2013)

Loading...


Adsorption and microstructure of core-shell nanoparticles at liquid-liquid interfaces: an X-ray reflectivity study. Isa L Chimia 67(4), 297, (2013)

Loading...


Low-temperature preparation of functional carbon nanocapsules via self-assembly and carbonization of hexayne amphiphiles. Schrettl S and Frauenrath H Chimia 67(6), 429, (2013)

Loading...


Merck 14,10039

Sax 6, 2735

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?