EMAIL THIS PAGE TO A FRIEND

270741 Sigma-Aldrich

Reagent Alcohol

CHROMASOLV®, for HPLC

Synonym: Ethyl alcohol, Reagent alcohol, RGA, denatured alcohol

Purchase

Properties

Related Categories Amber Glass Bottles, CHROMASOLV for HPLC, Fixing Reagents, HIS Select Supporting Products and Reagents, HIS-Select,
grade   CHROMASOLV®
  denatured
  for HPLC
contains   5% isopropyl alcohol as denaturant
  5% methyl alcohol as denaturant
impurities   ≤0.10% water
evapn. residue   <0.0005%
λ   H2O reference
UV absorption   λ: 205 nm Amax: 1.00
  λ: 210 nm Amax: 0.65
  λ: 220 nm Amax: 0.35
  λ: 255 nm Amax: 0.04
  λ: 300-400 nm Amax: 0.01

Description

Components

95 parts of specially denatured ethyl alcohol 3A, 200 proof, with 5 parts of isopropyl alcohol. Final concentrations are ~ 90% ethanol, ~ 5% methanol and ~ 5% isopropanol.

Packaging

1, 6×1, 2, 4×2, 4×4 L in glass bottle

Legal Information

CHROMASOLV is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH

Price and Availability

TruPAGE™ Pre-cast Polyacrylamide Gels


TruPAGE™ Pre-cast Polyacrylamide GelsTruPAGE Gels run true every time!

  • . 2-Year Shelf Life
  • . Superior Band Resolution
  • . Strong & Tear Resistant
  • . Higher Capacity Wells


Customers Also Viewed

Ethanol
34852Ethanol

CHROMASOLV®, absolute, for HPLC, ≥99.8%

reagent grade

Ethyl alcohol, Pure

200 proof, HPLC/spectrophotometric grade

Reagent Alcohol

anhydrous, ≤0.003% water

Reagent Alcohol

spectrophotometric grade

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Hazard Codes (Europe) 
Risk Statements (Europe) 
Safety Statements (Europe) 
16-36/37
RIDADR 
UN 1170C 3 / PGII
WGK Germany 
1
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Demonstration of an in vivo functional beta 3-adrenoceptor in man. Enocksson S, Shimizu M, Lönnqvist F, et al. J. Clin. Invest. 95(5), 2239-45, (1995)

Loading...


Cheminformatics analysis of assertions mined from literature that describe drug-induced liver injury in different species. Denis Fourches et al Chem. Res. Toxicol. 23, 171-83, (2010)

Loading...


Dissecting a complex chemical stress: chemogenomic profiling of plant hydrolysates. Skerker JM, Leon D, Price MN, et al. Mol. Syst. Biol. 9, 674, (2013)

Loading...


Comparative polygenic analysis of maximal ethanol accumulation capacity and tolerance to high ethanol levels of cell proliferation in yeast. Pais TM, Foulquié-Moreno MR, Hubmann G, et al. PLoS Genet. 9(6), e1003548, (2013)

Loading...


Ethanol metabolism and its effects on the intestinal epithelial barrier. Elamin EE, Masclee AA, Dekker J, et al. Nutr. Rev. 71(7), 483-99, (2013)

Loading...


HMDB: a knowledgebase for the human metabolome. Wishart DS, Knox C, Guo AC, et al. Nucleic Acids Res. 37(Database issue), D603-10, (2009)

Loading...


Reduced biocide susceptibility in Candida albicans biofilms. Jeniel E Nett et al Antimicrob. Agents Chemother. 52, 3411-3, (2008)

Loading...


Increased plasma phenylacetic acid in patients with end-stage renal failure inhibits iNOS expression. Jankowski J, van der Giet M, Jankowski V, et al. J. Clin. Invest. 112(2), 256-64, (2003)

Loading...


Ethanol effects on N-methyl-D-aspartate receptors in the bed nucleus of the stria terminalis. Wills TA and Winder DG Cold Spring Harb. Perspect. Med. 3(4), a012161, (2013)

Loading...


Karrikins discovered in smoke trigger Arabidopsis seed germination by a mechanism requiring gibberellic acid synthesis and light. Nelson DC Plant Physiol. 149(2), 863-73, (2009)

Loading...


Prediction of skeletal muscle and fat mass in patients with advanced cancer using a metabolomic approach. Stretch C, Eastman T, Mandal R, et al. J. Nutr. 142(1), 14-21, (2012)

Loading...


Chemocatalytic conversion of ethanol into butadiene and other bulk chemicals. Angelici C, Weckhuysen BM, and Bruijnincx PC ChemSusChem 6(9), 1595-614, (2013)

Loading...


Lead identification of conformationally restricted ?-lactam type combretastatin analogues: synthesis, antiproliferative activity and tubulin targeting effects. Miriam Carr et al Eur. J. Med. Chem. 45, 5752-66, (2010)

Loading...


Tissue distribution of cocaine methyl esterase and ethyl transferase activities: correlation with carboxylesterase protein. Dean RA, Zhang J, Brzezinski MR, et al. J. Pharmacol. Exp. Ther. 275(2), 965-71, (1995)

Loading...


Ethanol-TGFalpha-MEK signaling promotes growth of human hepatocellular carcinoma. Hennig M J. Surg. Res. 154(2), 187-95, (2009)

Loading...


Air to lung partition coefficients for volatile organic compounds and blood to lung partition coefficients for volatile organic compounds and drugs. Michael H Abraham et al Eur. J. Med. Chem. 43, 478-85, (2008)

Loading...


1H and (13)C NMR spectroscopic analysis of human saliva. Silwood CJ, Lynch E, Claxson AW, et al. J. Dent. Res. 81(6), 422-7, (2002)

Loading...


Detection of autosomal dominant polycystic kidney disease by NMR spectroscopic fingerprinting of urine. Gronwald W, Klein MS, Zeltner R, et al. Kidney Int. 79(11), 1244-53, (2011)

Loading...


Effects of prolonged ethanol intake and malnutrition on rat pancreas. López JM, Bombi JA, Valderrama R, et al. Gut 38(2), 285-92, (1996)

Loading...


Endogenous A1 adenosine receptor protects mice from acute ethanol-induced hepatotoxicity. Yang P, Wang Z, Zhan Y, et al. Toxicology 309, 100-6, (2013)

Loading...


Alcohol, tobacco, and hypertension. Friedman GD, Klatsky AL, and Siegelaub AB Hypertension 4(5 Pt 2), III143-50, (1982)

Loading...


Vital signs: alcohol poisoning deaths - United States, 2010-2012. Kanny D, Brewer RD, Mesnick JB, et al. MMWR Morb. Mortal. Wkly. Rep. 63(53), 1238-42, (2015)

Loading...


Single hepatocellular carcinoma smaller than 2 cm: are ethanol injection and radiofrequency ablation equally effective? Pompili M, De Matthaeis N, Saviano A, et al. Anticancer Res. 35(1), 325-32, (2015)

Loading...


Impact of local steroid or statin treatment of experimental temporomandibular joint arthritis on bone growth in young rats. Holwegner C, Reinhardt AL, Schmid MJ, et al. Am. J. Orthod. Dentofacial Orthop. 147(1), 80-8, (2015)

Loading...


Bisphenol A release from orthodontic adhesives and its correlation with the degree of conversion. Purushothaman D, Kailasam V, and Chitharanjan AB Am. J. Orthod. Dentofacial Orthop. 147(1), 29-36, (2015)

Loading...


A practical guide for diagnosis and treatment of arteriovenous malformations in the oral and maxillofacial region. Su LX, Fan XD, Zheng JW, et al. Chin. J. Dent. Res. 17(2), 85-9, (2014)

Loading...


High-dose alcohol induces reactive oxygen species-mediated apoptosis via PKC-β/p66Shc in mouse primary cardiomyocytes. Wang Y, Zhao J, Yang W, et al. Biochem. Biophys. Res. Commun. 456(2), 656-61, (2015)

Loading...


Most of UK is now out of step with rest of Europe on drink driving limits. Christie B BMJ 349, g7533, (2014)

Loading...


The effect of thalidomide on ethanol-induced gastric mucosal damage in mice: involvement of inflammatory cytokines and nitric oxide. Amirshahrokhi K and Khalili AR Chem. Biol. Interact. 225, 63-9, (2015)

Loading...


[10-year-old boy with blackish brown skin alterations]. Guarneri C, Tchernev G, Chokoeva AA, et al. Dtsch. Med. Wochenschr. 139(50), 2585-6, (2014)

Loading...


Density, ultrasound velocity, acoustic impedance, reflection and absorption coefficient determination of liquids via multiple reflection method. Hoche S, Hussein MA, and Becker T Ultrasonics 57, 65-71, (2015)

Loading...


Ethanol induced impairment of glucose metabolism involves alterations of GABAergic signaling in pancreatic β-cells. Wang S, Luo Y, Feng A, et al. Toxicology 326, 44-52, (2014)

Loading...


Dietary saturated lipids in alcoholic liver disease: new microbiota-targeting bullets? Tilg H and Gao B Gastroenterology 148(1), 16-9, (2015)

Loading...


Alteration of BRCA1 expression affects alcohol-induced transcription of RNA Pol III-dependent genes. Zhong Q, Shi G, Zhang Y, et al. Gene 556(1), 74-9, (2015)

Loading...


Androgen therapy reverses injuries caused by ethanol consumption in the prostate: testosterone as a possible target to ethanol-related disorders. Mendes LO, Scarano WR, Rochel-Maia SS, et al. Life Sci. 120, 22-30, (2015)

Loading...


Ethanol ablation of predominantly cystic thyroid nodules: evaluation of recurrence rate and factors related to recurrence. Suh CH, Baek JH, Ha EJ, et al. Clin. Radiol. 70(1), 42-7, (2015)

Loading...


Alcohol, one-carbon nutrient intake, and risk of colorectal cancer according to tumor methylation level of IGF2 differentially methylated region. Nishihara R, Wang M, Qian ZR, et al. Am. J. Clin. Nutr. 100(6), 1479-88, (2014)

Loading...


Demethyleneberberine, a natural mitochondria-targeted antioxidant, inhibits mitochondrial dysfunction, oxidative stress, and steatosis in alcoholic liver disease mouse model. Zhang P, Qiang X, Zhang M, et al. J. Pharmacol. Exp. Ther. 352(1), 139-47, (2015)

Loading...


Moderate and severe traumatic brain injury: effect of blood alcohol concentration on Glasgow Coma Scale score and relation to computed tomography findings. Rundhaug NP, Moen KG, Skandsen T, et al. J. Neurosurg. 122(1), 211-8, (2015)

Loading...


Non-(13)CO2 targeted breath tests: a feasibility study. Ruzsanyi V, Lederer W, Seger C, et al. J. Breath Res. 8(4), 046005, (2014)

Loading...


Czech mass methanol outbreak 2012: epidemiology, challenges and clinical features. Zakharov S, Pelclova D, Urban P, et al. Clin. Toxicol. (Phila.) 52(10), 1013-24, (2014)

Loading...


Evidence for α-helices in the large intracellular domain mediating modulation of the α1-glycine receptor by ethanol and Gβγ. Burgos CF, Castro PA, Mariqueo T, et al. J. Pharmacol. Exp. Ther. 352(1), 148-55, (2015)

Loading...


Modelling of the growth/no growth interface of Wallemia sebi and Eurotium herbariorum as a function of pH, aw and ethanol concentration. Deschuyffeleer N, Vermeulen A, Daelman J, et al. Int. J. Food Microbiol. 192, 77-85, (2015)

Loading...


Ethanol-induced alcohol dehydrogenase E (AdhE) potentiates pneumolysin in Streptococcus pneumoniae. Luong TT, Kim EH, Bak JP, et al. Infect. Immun. 83(1), 108-19, (2015)

Loading...


Growth/no growth models for Zygosaccharomyces rouxii associated with acidic, sweet intermediate moisture food products. Marvig CL, Kristiansen RM, and Nielsen DS Int. J. Food Microbiol. 192, 51-7, (2015)

Loading...


Exhaust and evaporative emissions from motorcycles fueled with ethanol gasoline blends. Li L, Ge Y, Wang M, et al. Sci. Total Environ. 502, 627-31, (2015)

Loading...


Homocysteine promotes intestinal fibrosis in rats with trinitrobenzene sulfonic acid-induced colitis. Ding H, Gan HZ, Fan WJ, et al. Dig. Dis. Sci. 60(2), 375-81, (2015)

Loading...


Gastroprotective effect of nymphayol isolated from Nymphaea stellata (Willd.) flowers: contribution of antioxidant, anti-inflammatory and anti-apoptotic activities. Antonisamy P, Subash-Babu P, Alshatwi AA, et al. Chem. Biol. Interact. 224, 157-63, (2014)

Loading...


Biofuels. Altered sterol composition renders yeast thermotolerant. Caspeta L, Chen Y, Ghiaci P, et al. Science 346(6205), 75-8, (2014)

Loading...


Biofuels. Engineering alcohol tolerance in yeast. Lam FH, Ghaderi A, Fink GR, et al. Science 346(6205), 71-5, (2014)

Loading...


Microbiology. How to survive being hot and inebriated. Cheng C and Kao KC Science 346(6205), 35-6, (2014)

Loading...


Alcohol fat clearing increases lymph node yield after surgery for colorectal cancer. Duldulao M, Booth C, Denham L, et al. Am. Surg. 80(10), 1054-8, (2014)

Loading...


Reply: To PMID 24246526. Jiang H and Zhang WX Am. J. Obstet. Gynecol. 212(1), 118-9, (2015)

Loading...


Pregnancy outcomes of interventional ultrasound sclerotherapy with 98% ethanol on women with hydrosalpinx. Kasius JC and Broekmans FJ Am. J. Obstet. Gynecol. 212(1), 118, (2015)

Loading...


Glycine and GABA(A) ultra-sensitive ethanol receptors as novel tools for alcohol and brain research. Naito A, Muchhala KH, Asatryan L, et al. Mol. Pharmacol. 86(6), 635-46, (2014)

Loading...


Supplementation of saturated long-chain fatty acids maintains intestinal eubiosis and reduces ethanol-induced liver injury in mice. Chen P, Torralba M, Tan J, et al. Gastroenterology 148(1), 203-214.e16, (2015)

Loading...


Sensitization of ethanol-induced place preference as a result of up-regulation of type 1 inositol 1,4,5-trisphosphate receptors in mouse nucleus accumbens. Kurokawa K, Mizuno K, and Ohkuma S J. Neurochem. 131(6), 836-47, (2014)

Loading...


Increased activity of the complement system in the liver of patients with alcoholic hepatitis. Shen H, French BA, Liu H, et al. Exp. Mol. Pathol. 97(3), 338-44, (2014)

Loading...


Renewable Energy. Cellulosic ethanol at last? Service RF Science 345(6201), 1111, (2014)

Loading...


Flocculation causes inhibitor tolerance in Saccharomyces cerevisiae for second-generation bioethanol production. Westman JO, Mapelli V, Taherzadeh MJ, et al. Appl. Environ. Microbiol. 80(22), 6908-18, (2014)

Loading...


Utilization of by-products derived from bioethanol production process for cost-effective production of lactic acid. Moon SK, Wee YJ, and Choi GW J. Ind. Microbiol. Biotechnol. 41(10), 1525-31, (2014)

Loading...


Urban overdose hotspots: a 12-month prospective study in Dublin ambulance services. Klimas J, O'Reilly M, Egan M, et al. Am. J. Emerg. Med. 32(10), 1168-73, (2014)

Loading...


Reinforcing properties and neurochemical response of ethanol within the posterior ventral tegmental area are enhanced in adulthood by periadolescent ethanol consumption. Toalston JE, Deehan GA, Hauser SR, et al. J. Pharmacol. Exp. Ther. 351(2), 317-26, (2014)

Loading...


Reduction of an E. coli O157:H7 and Salmonella composite on fresh strawberries by varying antimicrobial washes and vacuum perfusion. Gurtler JB, Bailey RB, Jin TZ, et al. Int. J. Food Microbiol. 189, 113-8, (2014)

Loading...


[Subcutaneous implantation type central venous port management in patients with malignant tumors effect of different antiseptic agents on central venous port-related infection]. Sato J, Kumagai M, Kato K, et al. Gan To Kagaku Ryoho. 41(8), 981-5, (2014)

Loading...


An alteration of the gut-liver axis drives pulmonary inflammation after intoxication and burn injury in mice. Chen MM, Zahs A, Brown MM, et al. Am. J. Physiol. Gastrointest. Liver Physiol. 307(7), G711-8, (2014)

Loading...


The effects of alcohol on coagulation in trauma patients: interpreting thrombelastography with caution. Howard BM, Kornblith LZ, Redick BJ, et al. J. Trauma Acute Care Surg. 77(6), 865-71; discussion 871-2, (2014)

Loading...


Elucidating and alleviating impacts of lignocellulose-derived microbial inhibitors on Clostridium beijerinckii during fermentation of Miscanthus giganteus to butanol. Zhang Y and Ezeji TC J. Ind. Microbiol. Biotechnol. 41(10), 1505-16, (2014)

Loading...


Continuous rhythm monitoring for ventricular arrhythmias after alcohol septal ablation for hypertrophic cardiomyopathy. Balt JC, Wijffels MC, Boersma LV, et al. Heart 100(23), 1865-70, (2014)

Loading...


Differentially co-expressed genes in postmortem prefrontal cortex of individuals with alcohol use disorders: influence on alcohol metabolism-related pathways. Zhang H, Wang F, Xu H, et al. Hum. Genet. 133(11), 1383-94, (2014)

Loading...


Pleiotrophin differentially regulates the rewarding and sedative effects of ethanol. Vicente-Rodríguez M, Pérez-García C, Ferrer-Alcón M, et al. J. Neurochem. 131(5), 688-95, (2014)

Loading...


Ethanol affects hepatitis C pathogenesis: humanized SCID Alb-uPA mouse model. Osna NA, Kharbanda KK, Sun Y, et al. Biochem. Biophys. Res. Commun. 450(1), 773-6, (2014)

Loading...


Plant science. Best practices for biofuels. Youngs H and Somerville C Science 344(6188), 1095-6, (2014)

Loading...


Genistein supplementation increases bone turnover but does not prevent alcohol-induced bone loss in male mice. Yang CS, Mercer KE, Alund AW, et al. Exp. Biol. Med. (Maywood.) 239(10), 1380-9, (2014)

Loading...


Bactericidal efficacy of alcohol solution in community hospital and health centers. Tavichakorntrakool R, Sungkeeree S, Saisud P, et al. J. Med. Assoc. Thai. 97 Suppl 4, S44-8, (2014)

Loading...


Alcohol inhalation. Le Foll B and Loheswaran G CMAJ 186(10), E399, (2014)

Loading...


Using dexmedetomidine as adjunctive therapy for patients with severe alcohol withdrawal syndrome: another piece of the puzzle. Savel RH and Kupfer Y Crit. Care Med. 42(5), 1298-9, (2014)

Loading...


Evaluation of mutagenic and genotoxic activities of lobeline and its modulation on genomic instability induced by ethanol. da Costa E Silva LD, Rodrigues LC, Dos Santos VR, et al. Life Sci. 103(2), 73-8, (2014)

Loading...


Copper rewired. Nature 508(7495), 150, (2014)

Loading...


[The effect of ethanol on the endocrynic system]. Wolszczak B, Zasimowicz E, and Juchniewicz J Pol. Merkur. Lekarski 36(211), 45-7, (2014)

Loading...


Marshaling the autonomic nervous system for treatment of atrial fibrillation. Rajendran PS, Buch E, and Shivkumar K J. Am. Coll. Cardiol. 63(18), 1902-3, (2014)

Loading...


Effect of chronic consumption of Piliostigma thonningii on activities of alanine aminotransferase and aspartate aminotransferase in serum and liver in Rattus novergicus. Awhin EP, Jeroh E, Anigboro AA, et al. Pak. J. Biol. Sci. 16(24), 2062-5, (2013)

Loading...


In vitro anti cancer activity of ethanol extract fractions of Aerva lanata L. Bhanot A, Sharma R, Singh S, et al. Pak. J. Biol. Sci. 16(22), 1612-7, (2013)

Loading...


[Sobering up agents influencing toxicodynamics of ethanol]. Golovko AI Biomed. Khim. 59(6), 604-21, (2013)

Loading...


Eurycoma longifolia in Radix for the treatment of ethanol-induced gastric lesion in rats. Qodriyah HM and Asmadi AY Pak. J. Biol. Sci. 16(23), 1815-8, (2013)

Loading...


Sub-acute effects of ethanol extract of Sarcocephalus latifolius root on some physiologically important electrolytes in serum of normal Wistar albino rats. Enemor VH and Okaka AN Pak. J. Biol. Sci. 16(23), 1811-4, (2013)

Loading...


Cerebroprotective effect of isolated harmine alkaloids extracts of seeds of Peganum harmala L. on sodium nitrite-induced hypoxia and ethanol-induced neurodegeneration in young mice. Biradar SM, Joshi H, and Tarak KC Pak. J. Biol. Sci. 16(23), 1687-97, (2013)

Loading...


[Postoperative pancreonecrosis prophylaxis in a gastric cancer surgery]. Vinnik IuA, Oleksenko VV, Efetova TS, et al. Klin. Khir. (12), 10-4, (2013)

Loading...


[Protective effect of isoorientin on alcohol-induced hepatic fibrosis in rats]. Chen YX, Huang QF, Lin X, et al. Zhongguo Zhong Yao Za Zhi 38(21), 3726-30, (2013)

Loading...


[Effect of Dendrobium officinale granule on long-term-alcohol-induced hypertension rats]. Lv GY, Xia CQ, Chen SH, et al. Zhongguo Zhong Yao Za Zhi 38(20), 3560-5, (2013)

Loading...


[Cyanamide-ethanol reaction induced shock: report of a case and literature review]. Kondo Y, Fuke C, Higa A, et al. Chudoku. Kenkyu. 26(4), 295-9, (2013)

Loading...


The dangerous professor. Kupferschmidt K Science 343(6170), 478-81, (2014)

Loading...


[Alterations of prooxidant-antioxidant system of rat liver at ethanol and tetracycline action]. Nedoshytko KhIu Ukr. Biokhim. Zh. 85(5), 154-62, (2013)

Loading...


[Substrate specificity of Cryptococcus albidus and Eupenicillium erubescens alpha-L-rhamnosidases]. Hudzenko OV and Varbanets' LD Ukr. Biokhim. Zh. 85(5), 17-26, (2013)

Loading...


[Effect of growth factors and some microelements on biosurfactant synthesis of Acinetobacter calcoaceticus IMV B-7241]. Pirog TP, Shevchuk TA, Mashchenko OIu, et al. Mikrobiol. Z. 75(5), 18-26, (2013)

Loading...


Radiologic findings and percutaneous treatment of a rare giant soft tissue hydatid cyst. Yucesoy C, Ozturk E, and Hekimoglu B JBR-BTR 96(5), 286-9, (2013)

Loading...


[Characteristics of toxigenic properties of Myrothecium cinctum (Corda) sacc., 1886 and Myrothecium commune pidopl., 1969]. Zaĭchenko AM, Tsyganenko ES, Andrienko EV, et al. Mikrobiol. Z. 75(6), 41-5, (2013)

Loading...


[Alternative type of fuel--biobutanol]. Tigunova EA, Shul'ga SM, and Blium IaB Tsitol. Genet. 47(6), 51-71, (2013)

Loading...


[Isolation and characterization of lactose-fermenting yeasts Candida kefyr]. Ianieva OD, Voronina HO, and Pidhors'kyĭ VS Tsitol. Genet. 47(6), 43-50, (2013)

Loading...


[Metabolic engineering of yeast Hansenula polymorpha for construction of efficient ethanol producers]. Dmitruk KV and Sibirnyĭ AA Tsitol. Genet. 47(6), 3-21, (2013)

Loading...


Management of recurrent corneal erosions: are we getting better? Mencucci R and Favuzza E Br. J. Ophthalmol. 98(2), 150-1, (2014)

Loading...


Nanoberries for topical delivery of antioxidants. Montanari J, Vera M, Mensi E, et al. J. Cosmet. Sci. 64(6), 469-81, (2013)

Loading...


[Remarkable improvement in dyspnea following bronchofiberscopic ethanol injection therapy -a case report]. Shimoda K, Shiraishi Y, Harano T, et al. Gan To Kagaku Ryoho. 40(12), 2339-41, (2013)

Loading...


The evaluation of the amebicidal activity of Eryngium planum extracts. Derda M, Thiem B, Budzianowski J, et al. Acta Pol. Pharm. 70(6), 1027-34, (2013)

Loading...


Membrane bioreactors' potential for ethanol and biogas production: a review. Ylitervo P, Akinbomia J, and Taherzadeha MJ Environ. Technol. 34(13-16), 1711-23, (2013)

Loading...


Pharmacokinetics of ethanol in the maternal-fetal unit. Zelner I and Koren G J. Popul. Ther. Clin. Pharmacol. 20(3), e259-65, (2013)

Loading...


Alcohol and oral squamous cell carcinoma. Feller L, Chandran R, Khammissa RA, et al. SADJ 68(4), 176-80, (2013)

Loading...


[Advantages and disadvantages of zink-ethanol-formaldehyde as a fixative for immunocytochemistry and confocal laser microscopy]. Korzhevskiĭ DE, Sukhorukova EG, Gilerovich EG, et al. Morfologiia. 143(2), 81-5, (2013)

Loading...


Alcohol consumption as a preventive factor for developing rheumatoid arthritis: a dose-response meta-analysis of prospective studies. Jin Z, Xiang C, Cai Q, et al. Ann. Rheum. Dis. 73(11), 1962-7, (2014)

Loading...


[Influence of pH on synthesis of Acinetobacter calcoaceticus IMV B-7241 biosurfactants]. Pirog TP, Antoniuk SI, Konon AD, et al. Mikrobiol. Z. 75(3), 32-40, (2013)

Loading...


[Preparation of dealcholized onetaxotere®, docetaxel injection formulation and its pharmacokinetics in rats]. Onishi K, Tsukiyama I, Katoh M, et al. Gan To Kagaku Ryoho. 40(7), 891-6, (2013)

Loading...


[Features of spatial orientation formation in posterity of female rats with liver experimental defeat in Morris water maze]. Vakhnin VA and Brukhin GV Ross. Fiziol. Zh. Im. I. M. Sechenova 99(4), 464-70, (2013)

Loading...


[Cloning, expression and characterization of alcohol dehydrogenase and aldehyde dehydrogenase from Bacillus pseudofirmus OF4]. Ju J, Ma N, Zhao R, et al. Wei Sheng Wu Xue Bao 53(4), 363-71, (2013)

Loading...


[Transcript profile of converting xylose and glucose to ethanol by Candida shehatae]. Xiong X, Cai P, Xu Y, et al. Wei Sheng Wu Xue Bao 53(4), 339-45, (2013)

Loading...


Impact of reformulated ethanol-gasoline blends on high-emitting vehicles. Schifter I, Díaz L, and González U Environ. Technol. 34(5-8), 911-22, (2013)

Loading...


[Alcoholism and baclofen: a strange French affair]. Nau JY Rev. Med. Suisse 9(390), 1286-7, (2013)

Loading...


[Transcatheter arterial embolization with ethanol for symptomatic polycystic kidney disease]. Sakuhara Y Nihon Jinzo Gakkai Shi 55(4), 549-50, (2013)

Loading...


[Investigation into drinking problem of patients who visited a general hospital in central and northern Okinawa]. Nakai M, Hotta H, Ootsuru T, et al. Nihon Arukoru. Yakubutsu Igakkai Zasshi 48(2), 145-52, (2013)

Loading...


[Research on the actual conditions of alcohol dependence, the harmful use of alcohol and the optimal treatment in gastroenterological inpatients]. Ohwaki Y, Morita N, Ogai Y, et al. Nihon Arukoru. Yakubutsu Igakkai Zasshi 48(2), 126-44, (2013)

Loading...


Signs of stress: deep symmetric T-wave inversions. Moore PK, Marzec LN, and Krantz MJ Am. J. Med. 126(8), 682-4, (2013)

Loading...


[Chemical constituents from leaves of Nelumbo nucifera]. Zhao XL, Wang ZM, Ma XJ, et al. Zhongguo Zhong Yao Za Zhi 38(5), 703-8, (2013)

Loading...


[Effect of univalent cations on synthesis of surfactants by Acinetobacter calcoaceticus IMV B-7241]. Pirog TP, Shevchuk TA, Antoniuk SI, et al. Mikrobiol. Z. 75(2), 10-20, (2013)

Loading...


[Metabolism features of bacteria resistant to high concentrations of chromate]. Smirnova GF and Podgorskiĭ VS Mikrobiol. Z. 75(2), 3-9, (2013)

Loading...


[Optimization of extraction process of Chrysanthemi Flos by multi-index orthogonal experiment]. Zhang XH, Li H, Qin KM, et al. Zhongguo Zhong Yao Za Zhi 38(6), 821-4, (2013)

Loading...


This month in Aerospace Medicine history. Dalitsch WW Aviat. Space. Environ. Med. 84(5), 544, (2013)

Loading...


Percutaneous treatment of a sacral metastasis with combined embolization, cryoablation, alcohol ablation and sacroplasty for local tumor and pain control. Lee JH, Stein M, and Roychowdhury S Interv. Neuroradiol. 19(2), 250-3, (2013)

Loading...


Stink bug feeding induces fluorescence in developing cotton bolls. Xia J, Mustafic A, Toews MD, et al. J. Biol. Eng. 5(1), 11, (2011)

Loading...


The many roles for fluorine in medicinal chemistry. William K Hagmann et al J. Med. Chem. 51, 4359-69, (2008)

Loading...


Reduced paxillin expression contributes to the antimetastatic effect of 4-hydroxycoumarin on B16-F10 melanoma cells. Marco A Velasco-Velázquez et al Cancer Cell Int. 8, 8, (2008)

Loading...


[Depression of serum esterase and lipoprotein lipase activities in acute and longitudinal actions of ethanol]. Sokolik VV, Chursina VS, Artemchuk AA, et al. Biomed. Khim. 52(1), 95-100, (2006)

Loading...


[Ethanol changes sensitivity of Kupffer cells to endotoxin]. Yamashina S, Ikejima K, Enomoto N, et al. Nihon Arukoru. Yakubutsu Igakkai Zasshi 38(5), 415-24, (2003)

Loading...


Phototocatalytic lithography of poly(propylene sulfide) block copolymers: toward high-throughput nanolithography for biomolecular arraying applications. Jane P. Bearinger et al Langmuir 25, 1238-44, (2009)

Loading...


The Japanese toxicogenomics project: application of toxicogenomics. Takeki Uehara et al Mol. Nutr. Food. Res. 54, 218-27, (2010)

Loading...


Synthesis of fatty acid ethyl esters in mammalian tissues after ethanol exposure: a systematic review of the literature. Zelner I, Matlow JN, Natekar A, et al. Drug Metab. Rev. 45(3), 277-99, (2013)

Loading...


The human serum metabolome. Psychogios N, Hau DD, Peng J, et al. PLoS ONE 6(2), e16957, (2011)

Loading...


Frontiers of yeast metabolic engineering: diversifying beyond ethanol and Saccharomyces. Liu L, Redden H, and Alper HS Curr. Opin. Biotechnol. 24(6), 1023-30, (2013)

Loading...


Regulation of eumelanin/pheomelanin synthesis and visible pigmentation in melanocytes by ligands of the melanocortin 1 receptor. Le Pape E, Wakamatsu K, Ito S, et al. Pigment Cell Melanoma Res. 21(4), 477-86, (2008)

Loading...


A novel brain penetrant NPS receptor antagonist, NCGC00185684, blocks alcohol-induced ERK-phosphorylation in the central amygdala and decreases operant alcohol self-administration in rats. Thorsell A, Tapocik JD, Liu K, et al. J. Neurosci. 33(24), 10132-42, (2013)

Loading...


Dimethylglycine accumulates in uremia and predicts elevated plasma homocysteine concentrations. McGregor DO, Dellow WJ, Lever M, et al. Kidney Int. 59(6), 2267-72, (2001)

Loading...


Ethanol, Zn2+ and insulin interact as progression factors to enhance DNA synthesis synergistically in the presence of Ca2+ and other cell cycle initiators in fibroblasts. Huang JS, She QB, Crilly KS, et al. Biochem. J. 346 Pt 1, 241-7, (2000)

Loading...


Optimal antimicrobial catheter lock solution, using different combinations of minocycline, EDTA, and 25-percent ethanol, rapidly eradicates organisms embedded in biofilm. Issam Raad et al Antimicrob. Agents Chemother. 51, 78-83, (2007)

Loading...


Plasma amino acid profile on nocturnal hemodialysis. Raj DS, Ouwendyk M, Francoeur R, et al. Blood Purif. 18(2), 97-102, (2000)

Loading...


Aspergillus fumigatus RasA regulates asexual development and cell wall integrity. Fortwendel JR Eukaryotic Cell 7(9), 1530-9, (2008)

Loading...


Accumulation of trimethylamine and trimethylamine-N-oxide in end-stage renal disease patients undergoing haemodialysis. Bain MA, Faull R, Fornasini G, et al. Nephrol. Dial. Transplant. 21(5), 1300-4, (2006)

Loading...


Hemodynamic and metabolic responses to interstitial angiotensin II in normal weight and obese men. Boschmann M, Adams F, Schaller K, et al. J. Hypertens. 24(6), 1165-71, (2006)

Loading...


Intravesical ethanol test: an ineffective measure of bladder hyperpermeability. Gordon Z, Parsons CL, and Monga M Urology 61(3), 555-7, (2003)

Loading...


Aggregated DNA in ethanol solution. Piskur J and Rupprecht A FEBS Lett. 375(3), 174-8, (1995)

Loading...


A single bout of exercise is followed by a prolonged decrease in the interstitial glucose concentration in skeletal muscle. Henriksson J and Knol M Acta Physiol. Scand. 185(4), 313-20, (2005)

Loading...


Percutaneous ethanol injection for liver metastases. Riemsma RP, Bala MM, Wolff R, et al. Cochrane Database Syst. Rev. 5, CD008717, (2013)

Loading...


Improving the serum D-xylose test for the identification of patients with small intestinal malabsorption. Ehrenpreis ED, Salvino M, and Craig RM J. Clin. Gastroenterol. 33(1), 36-40, (2001)

Loading...


Enhanced urinary excretion of cysteinyl leukotrienes in patients with acute alcohol intoxication. Uemura M, Lehmann WD, Schneider W, et al. Gastroenterology 118(6), 1140-8, (2000)

Loading...


Unchanged serum levels of advanced glycation endproducts in patients with liver disease. Butscheid M, Schäfer C, Brenner S, et al. Naunyn Schmiedebergs Arch. Pharmacol. 375(6), 401-6, (2007)

Loading...


Calculating virtual log P in the alkane/water system (log P(N)(alk)) and its derived parameters deltalog P(N)(oct-alk) and log D(pH)(alk). Giulia Caron et al J. Med. Chem. 48, 3269-79, (2005)

Loading...


Serum concentrations of asymmetric (ADMA) and symmetric (SDMA) dimethylarginine in patients with chronic kidney diseases. Fleck C, Schweitzer F, Karge E, et al. Clin. Chim. Acta 336(1-2), 1-12, (2003)

Loading...


Peroral amphotericin B polymer nanoparticles lead to comparable or superior in vivo antifungal activity to that of intravenous Ambisome® or Fungizone™ Jagdishbhai L. Italia et al PLoS ONE 6, e25744, (2011)

Loading...


Is the rodent maternal separation model a valid and effective model for studies on the early-life impact on ethanol consumption? Nylander I and Roman E Psychopharmacology 229(4), 555-69, (2013)

Loading...


Circulating tetrahydrobiopterin concentrations in patients with septic shock. Galley HF, Le Cras AE, Yassen K, et al. Br. J. Anaesth. 86(4), 578-80, (2001)

Loading...


Changes in astrocyte shape induced by sublytic concentrations of the cholesterol-dependent cytolysin pneumolysin still require pore-forming capacity. Förtsch C, Hupp S, Ma J, et al. Toxins (Basel.) 3(1), 43-62, (2011)

Loading...


Relationship between omega-3 fatty acids and plasma neuroactive steroids in alcoholism, depression and controls. Nieminen LR, Makino KK, Mehta N, et al. Prostaglandins Leukot. Essent. Fatty Acids 75(4-5), 309-14, (2006)

Loading...


Ethanol-induced in vitro invasion of breast cancer cells: the contribution of MMP-2 by fibroblasts. Aye MM, Ma C, Lin H, et al. Int. J. Cancer 112(5), 738-46, (2004)

Loading...


A theoretical study of the thermodynamics of microbial growth using Saccharomyces cerevisiae and a different free energy equation. Battley EH Q. Rev. Biol. 88(2), 69-96, (2013)

Loading...


Oral L-carnitine: metabolite formation and hemodialysis. Bain MA, Faull R, Milne RW, et al. Curr. Drug Metab. 7(7), 811-6, (2006)

Loading...


Combined bezafibrate and medroxyprogesterone acetate: potential novel therapy for acute myeloid leukaemia. Khanim FL PLoS ONE 4(12), e8147, (2009)

Loading...


Plasma pteridine concentrations in patients with chronic renal failure. Yokoyama K, Tajima M, Yoshida H, et al. Nephrol. Dial. Transplant. 17(6), 1032-6, (2002)

Loading...


Influence of alcohol on peripheral neurotransmitter function. Pohorecky LA Fed. Proc. 41(8), 2452-5, (1982)

Loading...


Convenient QSAR model for predicting the complexation of structurally diverse compounds with beta-cyclodextrins. Alfonso Pérez-Garrido et al Bioorg. Med. Chem. 17, 896-904, (2009)

Loading...


A gas chromatographic--mass spectrometric study of profiles of volatile metabolites in hepatic encephalopathy. Goldberg EM, Blendis LM, and Sandler S J. Chromatogr. A 226(2), 291-9, (1981)

Loading...


Effect of surfaces on amyloid fibril formation. Moores B, Drolle E, Attwood SJ, et al. PLoS ONE 6(10), e25954, (2011)

Loading...


Sulpho-conjugation of ethanol in humans in vivo and by individual sulphotransferase forms in vitro. Schneider H and Glatt H Biochem. J. 383(Pt. 3), 543-9, (2004)

Loading...


Plasma free amino acids and their metabolites in Taiwanese patients on hemodialysis and continuous ambulatory peritoneal dialysis. Chuang CK, Lin SP, Chen HH, et al. Clin. Chim. Acta 364(1-2), 209-16, (2006)

Loading...


Pseudo-Cushing's syndrome: the role of alcohol. Elias AN, Meshkinpour H, Valenta LJ, et al. J. Clin. Gastroenterol. 4(2), 137-9, (1982)

Loading...


The effect of the radio-protective agents ethanol, trimethylglycine, and beer on survival of X-ray-sterilized male Aedes aegypti. Rodriguez SD, Brar RK, Drake LL, et al. Parasit. Vectors 6, 211, (2013)

Loading...


Poly(hydroxyalkanoates)-based polymeric nanoparticles for drug delivery. Errico C, Bartoli C, Chiellini F, et al. J. Biomed. Biotechnol. 2009, 571702, (2009)

Loading...


Extractionless method for the simultaneous high-performance liquid chromatographic determination of urinary caffeine metabolites for N-acetyltransferase 2, cytochrome P450 1A2 and xanthine oxidase activity assessment. Nyéki A, Biollaz J, Kesselring UW, et al. J. Chromatogr. B. Biomed. Sci. Appl. 755(1-2), 73-84, (2001)

Loading...


Solvothermal fabrication of rubidium tungsten bronze for the absorption of near infrared light. Guo C, Yin S, Dong Q, et al. J. Nanosci. Nanotechnol. 13(5), 3236-9, (2013)

Loading...


A co-beneficial system using aquatic plants: bioethanol production from free-floating aquatic plants used for water purification. Soda S, Mishima D, Inoue D, et al. Water Sci. Technol. 67(11), 2637-44, (2013)

Loading...


Arp2 depletion inhibits sheet-like protrusions but not linear protrusions of fibroblasts and lymphocytes. Susan M. Nicholson-Dykstra et al Cell Motil. Cytoskeleton 65, 904-22, (2009)

Loading...


The role of capsaicin-sensitive afferents in autonomic dysreflexia in patients with spinal cord injury. Igawa Y, Satoh T, Mizusawa H, et al. BJU Int. 91(7), 637-41, (2003)

Loading...


Lewis acid-water/alcohol complexes as hydrogen atom donors in radical reactions. Povie G and Renaud P Chimia 67(4), 250-2, (2013)

Loading...


1H-NMR studies of cerebrospinal fluid: endogenous ethanol in patients with cervical myelopathy. Meshitsuka S, Morio Y, Nagashima H, et al. Clin. Chim. Acta 312(1-2), 25-30, (2001)

Loading...


Alcohol dependence: molecular and behavioral evidence. Trudell JR, Messing RO, Mayfield J, et al. Trends Pharmacol. Sci. 35(7), 317-23, (2014)

Loading...


Roles of heat shock factor 1 in neuronal response to fetal environmental risks and its relevance to brain disorders. Hashimoto-Torii K, Torii M, Fujimoto M, et al. Neuron 82(3), 560-72, (2014)

Loading...


The effect of polaprezinc on gastric mucosal protection in rats with ethanol-induced gastric mucosal damage: comparison study with rebamipide. Choi HS, Lim JY, Chun HJ, et al. Life Sci. 93(2-3), 69-77, (2013)

Loading...


High cerebral scyllo-inositol: a new marker of brain metabolism disturbances induced by chronic alcoholism. Viola A, Nicoli F, Denis B, et al. MAGMA 17(1), 47-61, (2004)

Loading...


Conserved single residue in the BK potassium channel required for activation by alcohol and intoxication in C. elegans. Davis SJ, Scott LL, Hu K, et al. J. Neurosci. 34(29), 9562-73, (2014)

Loading...


Different Contributions of Dopamine D1 and D2 Receptor Activity to Alcohol Potentiation of Brain Stimulation Reward in C57BL/6J and DBA/2J Mice. Fish EW, DiBerto JF, Krouse MC, et al. J. Pharmacol. Exp. Ther., (2014)

Loading...


Overexpression of the steroidogenic enzyme cytochrome P450 side chain cleavage in the ventral tegmental area increases 3α,5α-THP and reduces long-term operant ethanol self-administration. Cook JB, Werner DF, Maldonado-Devincci AM, et al. J. Neurosci. 34(17), 5824-34, (2014)

Loading...


Separation of ovotransferrin and ovomucoid from chicken egg white. Abeyrathne ED, Lee HY, and Ahn DU Poult. Sci. 93(4), 1010-7, (2014)

Loading...


Nonenzymatic sugar production from biomass using biomass-derived γ-valerolactone. Luterbacher JS, Rand JM, Alonso DM, et al. Science 343(6168), 277-80, (2014)

Loading...


Hepatopoietin Cn reduces ethanol-induced hepatoxicity via sphingosine kinase 1 and sphingosine 1-phosphate receptors. Liu Y, Saiyan S, Men TY, et al. J. Pathol. 230(4), 365-76, (2013)

Loading...


Effect of the structural characteristics of binary Pt-Ru and ternary Pt-Ru-M fuel cell catalysts on the activity of ethanol electrooxidation in acid medium. Antolini E ChemSusChem 6(6), 966-73, (2013)

Loading...


Prenatal alcohol exposure causes the over-expression of DHAND and EHAND by increasing histone H3K14 acetylation in C57 BL/6 mice. Zhang W, Peng C, Zheng M, et al. Toxicol. Lett. 228(3), 140-6, (2014)

Loading...


Regulation of pancreatic inflammation by connective tissue growth factor (CTGF/CCN2). Charrier A, Chen R, Kemper S, et al. Immunology 141(4), 564-76, (2014)

Loading...


Short- and long-term effects of acetaldehyde on plasma. Brecher AS and Adamu MT Alcohol 26(1), 49-53, (2002)

Loading...


Relationship between the concentration of ethanol and methanol in blood samples from Swedish drinking drivers. Jones AW and Löwinger H Forensic Sci. Int. 37(4), 277-85, (1988)

Loading...


The FXR agonist 6ECDCA reduces hepatic steatosis and oxidative stress induced by ethanol and low-protein diet in mice. Lívero FA, Stolf AM, Dreifuss AA, et al. Chem. Biol. Interact. 217, 19-27, (2014)

Loading...


Left ventricular systolic function following alcohol septal ablation for symptomatic hypertrophic cardiomyopathy. Moss TJ, Krantz MJ, Zipse MM, et al. Am. J. Cardiol. 113(8), 1401-4, (2014)

Loading...


Mechanisms involved in the inhibitory effect of chronic alcohol exposure on pancreatic acinar thiamin uptake. Srinivasan P, Subramanian VS, and Said HM Am. J. Physiol. Gastrointest. Liver Physiol. 306(7), G631-9, (2014)

Loading...


Is using ethanol-glycine irrigating fluid monitoring and 'good surgical practice' enough to prevent harmful absorption during transurethral resection of the prostate? Collins JW, Macdermott S, Bradbrook RA, et al. BJU Int. 97(6), 1247-51, (2006)

Loading...


Effects of oral administration of zinc and diiodohydroxyquinolein on plasma zinc levels of uremic patients. Paniagua R, Claure R, Amato D, et al. Nephron 69(2), 147-50, (1995)

Loading...


Temporal patterns of inflammatory gene expression in local tissues after banding or burdizzo castration in cattle. Pang W BMC Vet. Res. 5, 36, (2009)

Loading...


Statin treatment and diabetes affect myeloperoxidase activity in maintenance hemodialysis patients. Stenvinkel P, Rodríguez-Ayala E, Massy ZA, et al. Clin. J. Am. Soc. Nephrol. 1(2), 281-7, (2006)

Loading...


Ethanol disrupts intestinal epithelial tight junction integrity through intracellular calcium-mediated Rho/ROCK activation. Elamin E, Masclee A, Dekker J, et al. Am. J. Physiol. Gastrointest. Liver Physiol. 306(8), G677-85, (2014)

Loading...


Down-regulation of EBV-LMP1 radio-sensitizes nasal pharyngeal carcinoma cells via NF-κB regulated ATM expression. Ma X, Yang L, Xiao L, et al. PLoS ONE 6(11), e24647, (2011)

Loading...


Fetal alcohol exposure disrupts metabolic signaling in hypothalamic proopiomelanocortin neurons via a circadian mechanism in male mice. Agapito MA, Zhang C, Murugan S, et al. Endocrinology 155(7), 2578-88, (2014)

Loading...


[Direct metabolites of ethanol as biological markers of alcohol use: basic aspects and applications]. Thon N, Weinmann W, Yegles M, et al. Fortschr. Neurol. Psychiatr. 81(9), 493-502, (2013)

Loading...


Alcohol distribution in different postmortem body fluids. De Martinis BS, de Paula CM, Braga A, et al. Hum. Exp. Toxicol. 25(2), 93-7, (2006)

Loading...


Serum 7 alpha-hydroxy-4-cholesten-3-one and selenohomocholyltaurine (SeHCAT) whole body retention in the assessment of bile acid induced diarrhoea. Brydon WG, Nyhlin H, Eastwood MA, et al. Eur. J. Gastroenterol. Hepatol. 8(2), 117-23, (1996)

Loading...


Drinking drivers in Sweden who consume denatured alcohol preparations: an analytical-toxicological study. Jones AW, Lund M, and Andersson E J. Anal. Toxicol. 13(4), 199-203, (1989)

Loading...


Systematic review with meta-analysis: alcohol consumption and the risk of colorectal adenoma. Zhu JZ, Wang YM, Zhou QY, et al. Aliment. Pharmacol. Ther. 40(4), 325-37, (2014)

Loading...


Endoscopic ultrasound-guided ethanol ablation therapy for tumors. Zhang WY, Li ZS, and Jin ZD World J. Gastroenterol. 19(22), 3397-403, (2013)

Loading...


Development and validation of an LC-MS assay for finasteride and its application to prostate cancer prevention trial sample analysis. Chen X J. Chromatogr. Sci. 46(4), 356-61, (2008)

Loading...


Plasma histamine in patients with chronic renal failure and nephrotic syndrome. Gill DS, Fonseca VA, Barradas MA, et al. J. Clin. Pathol. 44(3), 243-5, (1991)

Loading...


Prevention of delirium in trauma patients: are we giving thiamine prophylaxis a fair chance? Blackmore C, Ouellet JF, Niven D, et al. Can. J. Surg. 57(2), 78-81, (2014)

Loading...


Determination of polyamines in hydrolysates of uremic plasma by high-performance cation-exchange column chromatography. Takagi T, Chung TG, and Saito A J. Chromatogr. A 272(2), 279-85, (1983)

Loading...


Epigenetic effects of ethanol on the liver and gastrointestinal system. Shukla SD and Lim RW Alcohol Res. 35(1), 47-55, (2013)

Loading...


Evaluation of gasoline-denatured ethanol as a carbon source for denitrification. Kazasi A, Boardman GD, and Bott CB Water Environ. Res. 85(6), 549-57, (2013)

Loading...


Chronic ethanol exposure during development: disturbances of breathing and adaptation. Dubois CJ, Kervern M, Naassila M, et al. Respir. Physiol. Neurobiol. 189(2), 250-60, (2013)

Loading...


Enzymatic hydrolysis of water hyacinth biomass for the production of ethanol: optimization of driving parameters. Ganguly A, Das S, Bhattacharya A, et al. Indian J. Exp. Biol. 51(7), 556-66, (2013)

Loading...


Antidiabetic activity of heart wood of Pterocarpus marsupium Roxb. and analysis of phytoconstituents. Mishra A, Srivastava R, Srivastava SP, et al. Indian J. Exp. Biol. 51(5), 363-74, (2013)

Loading...


Cardiovascular collapse and disseminated intravascular coagulation as complications of ethanol embolization of arteriovenous malformations in the upper lip: case report and literature review. Wang D, Su L, and Fan X J. Oral Maxillofac. Surg. 72(2), 346-51, (2014)

Loading...


Diagnostic characteristics and application of alcohol biomarkers. Topic A and Djukic M Clin. Lab. 59(3-4), 233-45, (2013)

Loading...


In vitro anti-oxidative activities and GC-MS analysis of various solvent extracts of Cassia singueana parts. Ibrahim MA, Koorbanally NA, and Islam MS Acta Pol. Pharm. 70(4), 709-19, (2013)

Loading...


Gastroprotective effect of an ethanolic extract from Neoglaziovia variegata (Arruda) Mez (Bromeliaceae) in rats and mice. Machado FD, Silva FV, Fernandes HB, et al. Z. Naturforsch. C 68(3-4), 97-107, (2013)

Loading...


Cerebral blood flow and oxygen consumption during ethanol withdrawal in the rat. Hemmingsen R, Barry DI, Hertz MM, et al. Brain Res. 173(2), 259-69, (1979)

Loading...


Assay of 6-gingerol in CO2 supercritical fluid extracts of ginger and evaluation of its sustained release from a transdermal delivery system across rat skin. Chen Y, Zhang C, Zhang M, et al. Pharmazie 69(7), 506-11, (2014)

Loading...


Angiographic and echocardiographic correlates of suitable septal perforators for alcohol septal ablation in hypertrophic obstructive cardiomyopathy. Chan W, Williams L, Kotowycz MA, et al. Can. J. Cardiol. 30(8), 912-9, (2014)

Loading...


Pedestrian traffic deaths among residents, visitors, and homeless persons--Clark County, Nevada, 2008-2011. Hickox KL, Williams N, Beck LF, et al. MMWR Morb. Mortal. Wkly. Rep. 63(28), 597-602, (2014)

Loading...


Hydrophilic organic/salt-containing aqueous two-phase solvent system for counter-current chromatography: a novel technique for separation of polar compounds. Liu D, Zou X, Gao M, et al. J. Chromatogr. A 1356, 157-62, (2014)

Loading...


Evaluation of hydrolysis and alcoholysis reactions in gas chromatography/mass spectrometry inlets. Ai G, Sun T, and Dong X J. Chromatogr. A 1356, 283-8, (2014)

Loading...


Right portal vein embolization with absolute ethanol in major hepatic resection for hepatobiliary malignancy. Sofue K, Arai Y, Shimada K, et al. Br. J. Surg. 101(9), 1122-8, (2014)

Loading...


Solvent, drying time, and substrate affect the responses of lone star ticks (Acari: Ixodidae) to the repellents deet and picaridin. Carroll JF, Carroll JF, Kramer M, et al. J. Med. Entomol. 51(3), 629-37, (2014)

Loading...


Percutaneous bone lesion ablation. Filippiadis DK, Tutton S, and Kelekis A Radiol. Med. 119(7), 462-9, (2014)

Loading...


Evaluation of bioactive components and antioxidant and anticancer properties of citrus wastes generated during bioethanol production. Im SJ, Kim JH, and Kim MY Nat. Prod. Commun. 9(4), 483-6, (2014)

Loading...


The inhibitory effect of ethanol on Sestrin3 in the pathogenesis of ethanol-induced liver injury. Kang X, Petyaykina K, Tao R, et al. Am. J. Physiol. Gastrointest. Liver Physiol. 307(1), G58-65, (2014)

Loading...


Selection of non-Saccharomyces yeast strains for reducing alcohol levels in wine by sugar respiration. Quirós M, Rojas V, Gonzalez R, et al. Int. J. Food Microbiol. 181, 85-91, (2014)

Loading...


Functional effects of alcohol withdrawal syndrome on peripheral sympathetic neurotransmission in vas deferens of adult rats. Bomfim GH, Verde LF, Frussa-Filho R, et al. Life Sci. 108(1), 34-43, (2014)

Loading...


Luteolin alleviates alcoholic liver disease induced by chronic and binge ethanol feeding in mice. Liu G, Zhang Y, Liu C, et al. J. Nutr. 144(7), 1009-15, (2014)

Loading...


Potent inhibition of alcohol self-administration in alcohol-preferring rats by a κ-opioid receptor antagonist. Cashman JR and Azar MR J. Pharmacol. Exp. Ther. 350(1), 171-80, (2014)

Loading...


Cytogenetic analysis of oral mucosa cells, induced by chlorhexidine, essential oils in ethanolic solution and triclosan mouthwashes. Ros-Llor I and Lopez-Jornet P Environ. Res. 132, 140-5, (2014)

Loading...


Salvianolic acid B protects against acute ethanol-induced liver injury through SIRT1-mediated deacetylation of p53 in rats. Li M, Lu Y, Hu Y, et al. Toxicol. Lett. 228(2), 67-74, (2014)

Loading...


Driving intoxicated: is hospital admission protective against legal ramifications? Edavettal M, Rogers F, and Rittenhouse K J. Trauma Acute Care Surg. 76(5), 1335, (2014)

Loading...


Protective effect of flavonoid extract from Chinese bayberry (Myrica rubra Sieb. et Zucc.) fruit on alcoholic liver oxidative injury in mice. Liu H, Qi X, Cao S, et al. J. Nat. Med. 68(3), 521-9, (2014)

Loading...


Renewable jet fuel. Kallio P, Pásztor A, Akhtar MK, et al. Curr. Opin. Biotechnol. 26, 50-5, (2014)

Loading...


Type 2 ryanodine receptors are highly sensitive to alcohol. Ye Y, Jian K, Jaggar JH, et al. FEBS Lett. 588(9), 1659-65, (2014)

Loading...


Ethanol infusion in the vein of Marshall leads to parasympathetic denervation of the human left atrium: implications for atrial fibrillation. Báez-Escudero JL, Keida T, Dave AS, et al. J. Am. Coll. Cardiol. 63(18), 1892-901, (2014)

Loading...


Scopoletin prevents alcohol-induced hepatic lipid accumulation by modulating the AMPK-SREBP pathway in diet-induced obese mice. Lee HI, Yun KW, Seo KI, et al. Metabolism 63(4), 593-601, (2014)

Loading...


MicroRNA-29b regulates ethanol-induced neuronal apoptosis in the developing cerebellum through SP1/RAX/PKR cascade. Qi Y, Zhang M, Li H, et al. J. Biol. Chem. 289(14), 10201-10, (2014)

Loading...


Lymph node revealing solutions in colorectal cancer: should they be used routinely? Horne J, Bateman AC, Carr NJ, et al. J. Clin. Pathol. 67(5), 383-8, (2014)

Loading...


Drugs and other chemicals involved in fatal poisoning in England and Wales during 2000 – 2011. Handley SA and Flanagan RJ Clin. Toxicol. (Phila.) 52(1), 1-12, (2014)

Loading...


In vitro antioxidant activities of leaf and root extracts of Albizia antunesiana harms. Chipiti T, Ibrahim MA, Koorbanally NA, et al. Acta Pol. Pharm. 70(6), 1035-43, (2013)

Loading...


A comparative study of low-fluence 1064-nm Q-switched Nd:YAG laser with or without chemical peeling using Jessner's solution in melasma patients. Lee DB, Suh HS, and Choi YS J. Dermatolog. Treat. 25(6), 523-8, (2014)

Loading...


Lignocellulosic ethanol production at high-gravity: challenges and perspectives. Koppram R, Tomás-Pejó E, Xiros C, et al. Trends Biotechnol. 32(1), 46-53, (2014)

Loading...


The B-vitamins in malnutrition with alcoholism. A model of intervitamin relationships. Dastur DK, Santhadevi N, Quadros EV, et al. Br. J. Nutr. 36(2), 143-59, (1976)

Loading...


Maltose and isomaltose in uremic plasma following icodextrin administration. Burke RA, Moberly JB, Patel H, et al. Adv. Perit. Dial. 14, 120-3, (1998)

Loading...


Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?