• USA Home
  • 27272 - α-Chymotrypsin from bovine pancreas

EMAIL THIS PAGE TO A FRIEND
27272 Sigma-Aldrich

α-Chymotrypsin from bovine pancreas

powder, almost white, ≥50 U/mg

Purchase

Properties

Related Categories 3.4.x.x Peptidases, 3.x.x.x Hydrolases, Application Index, Biochemicals and Reagents, Enzyme Class Index,
form   powder
mol wt   Mr ~25000(lit.)
  mol wt 25 kDa
color   almost white
storage temp.   −20°C
Gene Information   cow ... CTRB1(618826)

Description

Biochem/physiol Actions

A serine protease that hydrolyzes peptide bonds with aromatic or large hydrophobic side chains (Tyr, Trp, Phe, Met, Leu) on the carboxyl end of the bond.

Other Notes

Sales restrictions may apply

Unit Definition

1 U corresponds to the amount of enzyme, which hydrolyzes 1 μmol Suc-(Ala)2-Pro-Phe-4-NA per minute at pH 7.8 and 25°C

Price and Availability


Laboratory Gloves

Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
1
RTECS 
GC3050000

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C4129 α-Chymotrypsin from bovine pancreas, Type II, lyophilized powder, ≥40 units/mg protein

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?