EMAIL THIS PAGE TO A FRIEND
274364 Sigma-Aldrich

TWEEN® 80

average Mn ~1,310

Synonym: Polyoxyethylenesorbitan monooleate

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Culture, Detergents, Detergents A to M, Detergents by Applications,
vapor pressure   <1 mmHg ( 20 °C)
description   non-ionic
mol wt   average Mn ~1,310
refractive index   n20/D 1.472(lit.)
density   1.064 g/mL at 25 °C(lit.)
HLB   15

Description

Legal Information

TWEEN is a registered trademark of Croda International PLC

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
WG2932500
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?