• USA Home
  • 27545 - Clonidine hydrochloride

EMAIL THIS PAGE TO A FRIEND
27545 Sigma-Aldrich

Clonidine hydrochloride

≥98.0% (TLC)

Synonym: 2-(2,6-Dichloroanilino)-2-imidazoline hydrochloride

  • CAS Number 4205-91-8

  • Empirical Formula (Hill Notation) C9H9Cl2N3 · HCl

  • Molecular Weight 266.55

  •  Beilstein Registry Number 4163525

  •  EC Number 224-121-5

  •  MDL number MFCD00036705

  •  PubChem Substance ID 329753002

Purchase

Properties

InChI Key   ZNIFSRGNXRYGHF-UHFFFAOYSA-N
assay   ≥98.0% (TLC)
impurities   ≤1% water
solubility   H2O: 50 mg/mL clear, colorless
storage temp.   2-8°C

Description

Other Notes

Interacts with α-adrenergic receptors

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811PIH 6.1 / PGI
WGK Germany 
3
RTECS 
NJ2490000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C7897 Clonidine hydrochloride, solid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?