• USA Home
  • 27593 - Coenzyme A sodium salt hydrate

EMAIL THIS PAGE TO A FRIEND
27593 Sigma-Aldrich

Coenzyme A sodium salt hydrate

≥96% (HPLC)

Synonym: CoA Na2

  • CAS Number 55672-92-9 (anhydrous)

  • Empirical Formula (Hill Notation) C21H36N7O16P3S · xNa+ · yH2O

  • Molecular Weight 767.53 (anhydrous free acid basis)

  •  EC Number 259-747-8

Purchase

Properties

Related Categories Biochemicals and Reagents, Coenzyme A and Other Acyl Transfer Reagents, Cofactors, Enzymes, Inhibitors, and Substrates
description   cofactor for acyl transfer
assay   ≥96% (HPLC)
storage temp.   −20°C

Description

Other Notes

Review: Immobilized coenzymes and derivatives

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C3144 Coenzyme A sodium salt hydrate, cofactor for acyl transfer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?