EMAIL THIS PAGE TO A FRIEND
27600 Sigma-Aldrich

Caffeine

purum, anhydrous, ≥99.0% (HPLC)

Synonym: 1,3,7-Trimethylxanthine

  • CAS Number 58-08-2

  • Empirical Formula (Hill Notation) C8H10N4O2

  • Molecular Weight 194.19

  •  Beilstein Registry Number 17705

  •  EC Number 200-362-1

  •  MDL number MFCD00005758

Purchase

Properties

Related Categories Alkaloid, Cell Biology, Nutrition Research, Phytochemicals by Chemical Classification
InChI Key   RYYVLZVUVIJVGH-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (HPLC)
mp   233-238 °C
  234-236.5 °C(lit.)
Gene Information   human ... ADORA1(134), PDE4B(5142)

Description

Other Notes

Pharmacology and toxicology

Same product — same quality — now Sigma-Aldrich brand

Packaging

25, 100, 500 g in poly bottle

Biochem/physiol Actions

A central nervous system stimulant believed to act through adenosine receptors and monoamine neurotransmitters. It is an adenosine receptor antagonist and adenosine 3′,5′-cyclic monophosphate (cAMP) phosphodiesterase inhibitor. Thus, levels of cAMP increase in cells following treatment with caffeine. It has been reported to affect cellular calcium levels, releasing calcium from intracellular stores. It overrides the cell cycle effects of various chemicals such as protease inhibitors, preventing apoptosis; and it has been shown to inhibit cellular DNA repair mechanisms.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
EV6475000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?