276367 Sigma-Aldrich

Mercury(II) oxide

98%

DOWNLOAD MSDS (PDF)

Synonym: Mercuric oxide

  • CAS Number 21908-53-2

  • Empirical Formula (Hill Notation) HgO

  • Molecular Weight 216.59

  •  EC Number 244-654-7

  •  MDL number MFCD00011045

  •  PubChem Substance ID 24856705

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Materials Science, Mercury, Metal and Ceramic Science,
assay   98%
color   yellow
mp   500 °C (dec.)(lit.)

Description

Packaging

100, 500 g in poly bottle

Price and Availability

Customers Also Viewed

Mercury(II) oxide

99.999% trace metals basis

Mercury(II) oxide

puriss. p.a., ACS reagent, yellow, ≥99.0% (KT)

Mercury(II) oxide red

ACS reagent, ≥99.0%

Mercury(II) oxide yellow

ACS reagent, ≥99.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?