• USA Home
  • 27669 - Collagenase Chromophore-Substrate Kit

EMAIL THIS PAGE TO A FRIEND
27669 Sigma-Aldrich

Collagenase Chromophore-Substrate Kit

for quantitative collagenase determination

Purchase

Properties

Related Categories Biochemicals and Reagents, Collagenase Substrates, Enzyme Substrates, Enzymes, Inhibitors, and Substrates, Substrates by Enzyme More...
shipped in   wet ice
storage temp.   −20°C

Description

Packaging

Set with 100 mg component A (Cat. No. 27667) and 50 mg component B (Cat. No. 27668)

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?