• USA Home
  • 27671 - Collagenase Chromophore-Substrate Kit (for quantitative Collagenase-Determination)

EMAIL THIS PAGE TO A FRIEND
27671 Sigma-Aldrich

Collagenase Chromophore-Substrate Kit (for quantitative Collagenase-Determination)

Purchase

Properties

Related Categories Biochemicals and Reagents, Collagenase Substrates, Enzyme Substrates, Enzymes, Inhibitors, and Substrates, Substrates by Enzyme More...
shipped in   wet ice
storage temp.   −20°C

Description

Other Notes

Substrate for the spectrophotometric determination of collagenase

Packaging

Set with 1 g component A (Cat. No. 27667) and 500 mg component B (Cat. No. 27668)

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?