• USA Home
  • 27692 - 2,4,6-Trimethylpyridine

EMAIL THIS PAGE TO A FRIEND
27692 Sigma-Aldrich

2,4,6-Trimethylpyridine

≥98.0% (GC)

Synonym: 2,4,6-Collidine, sym-Collidine

  • CAS Number 108-75-8

  • Empirical Formula (Hill Notation) C8H11N

  • Molecular Weight 121.18

  •  Beilstein Registry Number 107283

  •  EC Number 203-613-3

  •  MDL number MFCD00006338

  •  PubChem Substance ID 329753014

Purchase

Properties

Related Categories Building Blocks, C8, Chemical Synthesis, Heterocyclic Building Blocks, Pyridines More...
InChI Key   BWZVCCNYKMEVEX-UHFFFAOYSA-N
assay   ≥98.0% (GC)
refractive index   n20/D 1.498(lit.)
  n20/D 1.498
bp   171-172 °C(lit.)
mp   −43 °C(lit.)
density   0.914 g/mL at 20 °C
  0.917 g/mL at 25 °C(lit.)

Description

Application

Tissue fixative for electron microscopy.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
RIDADR 
UN 1992 6.1(3) / PGIII
WGK Germany 
3
RTECS 
UU0970000
Flash Point(F) 
131 °F
Flash Point(C) 
55 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?