EMAIL THIS PAGE TO A FRIEND
27696 Sigma-Aldrich

Mevastatin

≥96% (HPLC)

Synonym: Compactin, ML-236B

  • CAS Number 73573-88-3

  • Empirical Formula (Hill Notation) C23H34O5

  • Molecular Weight 390.51

  •  Beilstein Registry Number 1269441

  •  MDL number MFCD05662341

Purchase

Properties

Related Categories Antibacterial, Antibiotics, Antibiotics A to Z, Antibiotics G-M, Biochemicals and Reagents,
InChI Key   AJLFOPYRIVGYMJ-INTXDZFKSA-N
assay   ≥96% (HPLC)
mp   151-153 °C
storage temp.   2-8°C

Description

Other Notes

Potent inhibitor of 3-hydroxy-3-methylglutaryl-CoA reductase, HMG-CoA reductase; Used e.g. in culturing compactin-resistant cell lines overaccumulating HMG-CoA reductase

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?