• USA Home
  • 27791 - Controlled Pore Glass, aminopropyl modified

EMAIL THIS PAGE TO A FRIEND
27791 Sigma-Aldrich

Controlled Pore Glass, aminopropyl modified

170 Å, extent of labeling: 0.2 mmol/g loading

Synonym: Aminopropyl-CPG

Purchase

Properties

Related Categories Aminopropyl, Analytical/Chromatography, Anion, LPLC Separation Media, LPLC/TLC/Paper Chromatography More...
extent of labeling   0.2 mmol/g loading
particle size   120-200 mesh

Description

Other Notes

1 EA contains 10 g

Application

Controlled Pore Glass, aminopropyl modified may be used in separation media for TLC, paper and LPL chromatography.

General description

Controlled pore glass is used as multipurpose support material since it possess high surface area and loading capacity, low relative cost, ease of surface derivatization, high--pressure applicability, and an invariable, rigid structure that withstands most solvent systems. It is used as immobilization matrix in packed-bed reactors. The glass beads undergo aminopropylation in nitric acid and (3-aminopropyltriethoxysilane) APES solution to form aminopropyl modified CPG.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?