EMAIL THIS PAGE TO A FRIEND
27963 Sigma-Aldrich

Kolliphor® EL

Synonym: Cremophor® EL, Macrogolglycerol ricinoleate, PEG-35 castor oil, Polyoxyl 35 hydrogenated castor oil, Polyoxyl-35 castor oil

Purchase

Properties

Related Categories Biochemicals and Reagents, Detergents, Detergents A to M, Non-Ionic Detergents
description   non-ionic
density   1.05 g/mL at 20 °C

Description

Application

Used to emulsify and solubilize oils and other water-insoluble substances. Cremophor is the brand name for a range of non-ionic polyethoxylated detergents. It was originally developed for use as solubilizers and emulsifiers.
This research grade product is intended for use in R&D and development only.

Other Notes

A derivative of castor oil and ethylene oxide.

This product has been replaced by C5135-SIGMA | Kolliphor EL pH-range 6.0 - 8.0

Legal Information

Cremophor is a registered trademark of BASF SE

Kolliphor is a registered trademark of BASF

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
Flash Point(F) 
494.6 °F
Flash Point(C) 
257 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C5135 Kolliphor® EL, pH-range 6.0 - 8.0

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?