EMAIL THIS PAGE TO A FRIEND
28051 Sigma-Aldrich

Crotonoyl chloride

technical, ≥90% (GC)

  • CAS Number 625-35-4

  • Linear Formula CH3CH=CHCOCl

  • Molecular Weight 104.53

  •  Beilstein Registry Number 506118

  •  EC Number 210-889-9

  •  MDL number MFCD00000734

  •  PubChem Substance ID 24856945

Purchase

Properties

Related Categories Acid Halides, Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks More...
InChI Key   RJUIDDKTATZJFE-NSCUHMNNSA-N
grade   technical
assay   ≥90% (GC)
contains   hydroquinone as stabilizer
refractive index   n20/D 1.459(lit.)
  n20/D 1.459
bp   120-123 °C(lit.)
density   1.08 g/mL at 20 °C
  1.091 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2920 3(8) / PGII
WGK Germany 
3
Flash Point(F) 
95 °F
Flash Point(C) 
35 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?