• USA Home
  • 28091 - Crotyl alcohol, mixture of cis and trans

EMAIL THIS PAGE TO A FRIEND
28091 Sigma-Aldrich

Crotyl alcohol, mixture of cis and trans

purum, mixture of cis- and trans-isomers (~1:19), ≥97.0% (GC)

Synonym: 2-Buten-1-ol

  • CAS Number 6117-91-5

  • Linear Formula CH3CH=CHCH2OH

  • Molecular Weight 72.11

  •  Beilstein Registry Number 1361395

  •  EC Number 228-086-7

  •  MDL number MFCD00002923

  •  PubChem Substance ID 24856958

Purchase

Properties

Related Categories Acyclic, Alkenes, Building Blocks, Chemical Synthesis, Organic Building Blocks More...
InChI Key   WCASXYBKJHWFMY-NSCUHMNNSA-N
grade   purum
assay   ≥97.0% (GC)
impurities   ~1% water
  ~2% butanol
refractive index   n20/D 1.427(lit.)
  n20/D 1.427
bp   121-122 °C(lit.)
density   0.845 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1987C 3 / PGIII
WGK Germany 
3
RTECS 
EM9275000
Flash Point(F) 
93.2 °F
Flash Point(C) 
34 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

C86006 Crotyl alcohol, mixture of cis and trans, 96%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?