EMAIL THIS PAGE TO A FRIEND
28123 Sigma-Aldrich

15-Crown-5

purum, ≥98.0% (GC)

Synonym: 1,4,7,10,13-Pentaoxacyclopentadecane

  • CAS Number 33100-27-5

  • Empirical Formula (Hill Notation) C10H20O5

  • Molecular Weight 220.26

  •  Beilstein Registry Number 1618144

  •  EC Number 251-379-6

  •  MDL number MFCD00005110

  •  PubChem Substance ID 329753050

Purchase

Properties

Related Categories Chelation/Complexation Compounds, Chemical Synthesis, Crown Ethers, Synthetic Reagents
InChI Key   VFTFKUDGYRBSAL-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.465(lit.)
  n20/D 1.465
bp   93-96 °C/0.05 mmHg(lit.)
density   1.113 g/mL at 20 °C(lit.)

Description

Other Notes

Phase transfer catalyst, complexing-agent

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
RTECS 
SB0200000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

188832 15-Crown-5, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?