EMAIL THIS PAGE TO A FRIEND
28202 Sigma-Aldrich

imMedia Amp Blue

for microbiology

Purchase

Properties

Related Categories All Liquid Media/Broths, All Solid Media/Agars, Bacterial Culture Media, Media, Microbial Media for Molecular Biology and Protein Expression,
grade   for microbiology
shelf life   limited shelf life, expiry date on the label

Description

Application

E. coli growth agar with ampicillin, IPTG and X-Gal, ready to use. Specially formulated growth medium for E. coli that eliminates the time consuming need for autoclaving in media preparation. It is ready to use after heating for only approx. 4 min. in a microwave oven.
Contents: 20 pouches. Each pouch provides sufficient material for 200 mL E. coli growth medium (8 to 10 plates). Presterilized low salt LB agar medium supplemented with ampicillin, IPTG and X-Gal for the blue/white recombinant E. coli screening.

Legal Information

imMedia is a trademark of Invitrogen Corp.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the environment, microbes have adapted to the habitats most suitable for thei...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?