EMAIL THIS PAGE TO A FRIEND
28222 Sigma-Aldrich

Chromoionophore X

Selectophore

Synonym: 4-Dioctylamino-4′-(trifluoroacetyl)stilbene, ETH 4004

Purchase

Properties

Related Categories Analytical/Chromatography, Chromoionophores, Ion Sensor Materials More...
grade   Selectophore
InChI Key   LQCOAQAROPQVJL-FOCLMDBBSA-N

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Chromoionophore for the detection of alcohol

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

27086 Chromoionophore I, Selectophore
27087 Chromoionophore II, Selectophore
27088 Chromoionophore III, Selectophore
27089 Chromoionophore IV, Selectophore
27095 Chromoionophore VI, Selectophore
27097 Chromoionophore VII, Selectophore
27098 Chromoionophore VIII, Selectophore
27102 Chromoionophore XI, Selectophore
28218 Chromoionophore IX, Selectophore
19684 Chromoionophore XVII, Selectophore

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?