• USA Home
  • 28242 - Potassium tetrakis(4-biphenylyl)borate

EMAIL THIS PAGE TO A FRIEND
28242 Sigma-Aldrich

Potassium tetrakis(4-biphenylyl)borate

≥97.0%

Synonym: Tetrakis(4-biphenylyl)boron potassium

Purchase

Properties

Related Categories Additives for ISE, Analytical/Chromatography, Ion Sensor Materials More...
InChI Key   FMJZZAIWRYCJQG-UHFFFAOYSA-N
assay   ≥97.0%
  >97.0% (CH)

Description

General description

Visit our Sensor Applications portal to learn more.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?