EMAIL THIS PAGE TO A FRIEND
28250 Sigma-Aldrich

Cumene hydroperoxide

technical, ~80% in cumene

Synonym: α,α-Dimethylbenzyl hydroperoxide

  • CAS Number 80-15-9

  • Linear Formula C6H5C(CH3)2OOH

  • Molecular Weight 152.19

  •  Beilstein Registry Number 1908117

  •  MDL number MFCD00002129

  •  PubChem Substance ID 329753108

Purchase

Properties

vapor density   5.4 (vs air)
vapor pressure   <0.03 mmHg ( 20 °C)
InChI Key   YQHLDYVWEZKEOX-UHFFFAOYSA-N
grade   technical
concentration   ~80% in cumene
refractive index   n20/D 1.519
bp   100-101 °C/8 mmHg(lit.)
density   1.028 g/mL at 20 °C
storage temp.   2-8°C

Description

Other Notes

Sales restrictions may apply

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
RIDADR 
UN 3109A3 8(5.2) / PGII
WGK Germany 
2
Flash Point(F) 
127.4 °F
Flash Point(C) 
53 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

247502 Cumene hydroperoxide, technical grade, 80%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?