• USA Home
  • 283940 - Rhodamine 123 hydrate

EMAIL THIS PAGE TO A FRIEND
283940 Sigma-Aldrich

Rhodamine 123 hydrate

Dye content 95 %

Purchase

Properties

InChI Key   SEXIURXGNASJRB-UHFFFAOYSA-N
composition   Dye content, 95%
absorption   λmax 501 nm

Description

Features and Benefits

Mitochondrial specific vital stain

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

R8004 Rhodamine 123, mitochondrial specific fluorescent dye

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?