EMAIL THIS PAGE TO A FRIEND
28460 Sigma-Aldrich

Methyl cyanoacetate

purum, ≥99.0% (GC)

  • CAS Number 105-34-0

  • Linear Formula NCCH2COOCH3

  • Molecular Weight 99.09

  •  Beilstein Registry Number 773945

  •  EC Number 203-288-8

  •  MDL number MFCD00001939

Purchase

Properties

Related Categories Building Blocks, C2 to C5, Carbonyl Compounds, Chemical Synthesis, Esters,
vapor density   3.41 (vs air)
vapor pressure   0.2 mmHg ( 20 °C)
InChI Key   ANGDWNBGPBMQHW-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
refractive index   n20/D 1.417(lit.)
  n20/D 1.418
bp   204-207 °C(lit.)
mp   −13 °C(lit.)
density   1.123 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
AG4375000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

108421 Methyl cyanoacetate, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?