• USA Home
  • 28480 - α-Cyano-4-hydroxycinnamic acid

EMAIL THIS PAGE TO A FRIEND
28480 Sigma-Aldrich

α-Cyano-4-hydroxycinnamic acid

purum, ≥99.0% (HPLC)

Synonym: α-CCA, α-CHCA, α-Cyano, 4-HCCA, ACCA, alpha-Cyano-4-hydroxycinnamic acid

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Building Blocks, C10, Carbonyl Compounds,
InChI Key   AFVLVVWMAFSXCK-VMPITWQZSA-N
grade   purum
assay   ≥99.0% (HPLC)
mp   235-250 °C
  245-250 °C(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

476870 α-Cyano-4-hydroxycinnamic acid, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?