EMAIL THIS PAGE TO A FRIEND
28640 Sigma-Aldrich

Cyanuric acid

purum, anhydrous, ≥98.0% (T)

Synonym: 1,3,5-Triazine-2,4,6-triol, 2,4,6-Trihydroxy-1,3,5-triazine

  • CAS Number 108-80-5

  • Empirical Formula (Hill Notation) C3H3N3O3

  • Molecular Weight 129.07

  •  Beilstein Registry Number 126982

  •  EC Number 203-618-0

  •  MDL number MFCD00082990

  •  PubChem Substance ID 329753281

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Triazines
InChI Key   ZFSLODLOARCGLH-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (T)
mp   >360 °C(lit.)
solubility   0.1 M NaOH: 0.2 g/10 mL clear, colorless

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
XZ1800000

Documents

Certificate of Analysis

Protocols & Articles

Articles

LC/MS Analysis of Cyanuric Acid on Ascentis® Express HILIC

From our library of Articles, Sigma-Aldrich presents LC/MS Analysis of Cyanuric Acid on Ascentis® Express HILIC
Keywords: Chromatography, Environmental, Environmental Analysis, Herbicides, High performance liquid chromatography, Mass spectrometry, Pesticides

US FDA LIB 4423: GC Analysis of Melamine and Related Compounds in Dog Food (Spiked) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US FDA LIB 4423: GC Analysis of Melamine and Related Compounds in Dog Food (Spiked) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US FDA LIB 4423: GC Analysis of Melamine and Related Compounds in Dog Food on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US FDA LIB 4423: GC Analysis of Melamine and Related Compounds in Dog Food on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US FDA LIB 4423: GC Analysis of Melamine and Related Compounds on SLB®-5ms, Method Blank

From our library of Articles, Sigma-Aldrich presents US FDA LIB 4423: GC Analysis of Melamine and Related Compounds on SLB®-5ms, Method Blank
Keywords: Chromatography, Gas chromatography, Purification

US FDA LIB 4423: GC Analysis of Melamine and Related Compounds on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US FDA LIB 4423: GC Analysis of Melamine and Related Compounds on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?