• USA Home
  • 28672 - Aluminum chloride - ethanol solution

EMAIL THIS PAGE TO A FRIEND
28672 Sigma-Aldrich

Aluminum chloride - ethanol solution

~11% in ethanol/water, for TLC derivatization

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Derivatization Reagents, Derivatization Reagents TLC, Sprays,
grade   for TLC derivatization
InChI Key   VSCWAEJMTAWNJL-UHFFFAOYSA-K
concentration   ~11% in ethanol/water
density   0.944 g/mL at 20 °C
suitability   corresponds for TLC
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Target organs 
Central nervous system, Lungs
RIDADR 
UN 2924 8(3) / PGII
WGK Germany 
1
Flash Point(F) 
62.6 °F
Flash Point(C) 
17 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?