• USA Home
  • 28707 - β-Cyclodextrin hydrate

EMAIL THIS PAGE TO A FRIEND
28707 Sigma-Aldrich

β-Cyclodextrin hydrate

puriss., ≥99.0% (HPLC)

Synonym: β-Schardinger dextrin, Cycloheptaamylose, Schardinger β-Dextrin hydrate

  • CAS Number 68168-23-0

  • Empirical Formula (Hill Notation) C42H70O35 · xH2O

  • Molecular Weight 1134.98 (anhydrous basis)

  •  EC Number 231-493-2

  •  MDL number MFCD00150811

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Capillary Electrophoresis, Chelation/Complexation Compounds, Chemical Synthesis,
InChI Key   JNSAKCOAFBFODP-ZQOBQRRWSA-N
grade   puriss.
assay   ≥99.0% (HPLC)
optical activity   [α]20/D +142±3°, c = 1% in H2O
loss   10-13% loss on drying
mp   >260 °C (dec.)(lit.)

Description

Other Notes

Cyclodextrins are low molecular weight organic molecules forming water-soluble complexes with various compounds

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?