EMAIL THIS PAGE TO A FRIEND
288551 Sigma-Aldrich

Silica gel

TLC high-purity grade, 5-25 μm, pore size 60 Å, with gypsum binder and fluorescent indicator, pore volume 0.75 cm3/g

Purchase

Properties

Related Categories Adsorbents for TLC, Analytical/Chromatography, LPLC/TLC/Paper Chromatography, Silica TLC Adsorbent, with fluorescent indicator, with binder, Silica TLC Adsorbents,
grade   TLC high-purity grade
description   Fe3+ ≤0.001%
InChI Key   VYPSYNLAJGMNEJ-UHFFFAOYSA-N
form   powder
contains   10% gypsum as binder
particle size   5-25 μm
pore size   0.75 cm3/g pore volume
  60 Å
surface area   BET surf. area ~500 m2/g
pH   ~6.8 (10% in aq. suspension)
bp   2230 °C
mp   >1600 °C
anion traces   chloride (Cl-): ≤0.003%

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
VV7315000
Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

288586 Silica gel, high-purity grade, pore size 60 Å, 2-25 μm particle size, without binder, with fluorescent indicator, pore volume 0.75 cm3/g, for thin layer chromatography

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?